Merck
Diazinon and permethrin mitigation across a grass-wetland buffer.
Bulletin of environmental contamination and toxicology (2014-08-26)
M T Moore, R Kröger, M A Locke, R E Lizotte, S Testa, C M Cooper
ABSTRACT
Vegetated buffers of different designs are often used as edge-of-field treatment practices to remove pesticides that may be entrained in agricultural runoff. However, buffer system efficacy in pesticide runoff mitigation varies widely due to a multitude of factors including, but not limited to, pesticide chemistry, vegetation composition, and hydrology. Two experimental systems, a control (no vegetation) and a grass-wetland buffer system, were evaluated for their ability to retain diazinon and permethrin associated with a simulated storm runoff. The two systems were equally inefficient at retaining diazinon (mean 9.6 % retention for control and buffer). Grass-wetland buffers retained 83 % and 85 % of cis- and trans-permethrin masses, respectively, while the control only retained 39 % and 44 % of cis- and trans-permethrin masses, respectively. Half-distances (the distance required to decrease pesticide concentration by one-half) for both permethrin isomers were 26 %-30 % shorter in grass buffers (22-23 m) than in the control (32 m). The current study demonstrates treatment efficacy was a function of pesticide properties with the more strongly sorbing permethrin retained to a greater degree. The study also demonstrates challenges in remediating multiple pesticides with a single management practice. By using suites of management practices, especially those employing vegetation, better mitigation of pesticide impacts may be accomplished.
Product Description
Supelco
Diazinon, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
Permethrin, PESTANAL®, analytical standard