Merck
  • Using the hemoglobin content of reticulocytes (RET-He) to evaluate anemia in patients with cancer.
Using the hemoglobin content of reticulocytes (RET-He) to evaluate anemia in patients with cancer.
American journal of clinical pathology (2014-09-23)
Ellinor I B Peerschke, Melissa S Pessin, Peter Maslak
ABSTRACT
Evaluation of anemia, particularly iron deficiency, in patients with cancer is difficult. This study examined using the hemoglobin content of reticulocytes (RET-He) to rule out iron deficiency, as defined by serum iron studies (transferrin saturation <20%, serum iron <40 μg/dL, and ferritin <100 ng/mL), in an unselected cancer patient population. Patients were entered into the study based on the existence of concurrent laboratory test requests for CBC and serum iron studies. Using a threshold of 32 pg/cell, RET-He ruled out iron deficiency with a negative predictive value (NPV) of 98.5% and 100%, respectively, in the study population (n = 209) and in a subpopulation of patients with low reticulocyte counts (n = 19). In comparison, the NPV of traditional CBC parameters (hemoglobin, <11 g/dL; mean corpuscular volume, <80 fL) was only 88.5%. These results support the use of RET-He in the evaluation of iron deficiency in a cancer care setting.
Sigma-Aldrich
Iron, puriss. p.a., carbonyl-Iron powder, low in magnesium and manganese compounds, ≥99.5% (RT)
Iron, IRMM®, certified reference material, 0.5 mm wire
Sigma-Aldrich
Iron, foil, thickness 0.1 mm, ≥99.9% trace metals basis
Sigma-Aldrich
Iron, chips, 99.98% trace metals basis
Sigma-Aldrich
Ferritin from equine spleen, Type I, saline solution
Iron, wire reel, 5m, diameter 0.025mm, as drawn, 99.99+%
Iron, wire reel, 1m, diameter 0.5mm, as drawn, 99.998%
Iron, wire reel, 0.5m, diameter 0.25mm, hard, 99.99+%
Iron, wire reel, 0.5m, diameter 1.0mm, as drawn, 99.99+%
Iron, wire reel, 5m, diameter 0.25mm, hard, 99.99+%
Iron, wire reel, 50m, diameter 0.5mm, as drawn, 99.95%
Iron, wire reel, 2m, diameter 0.25mm, annealed, 99.99+%
Iron, wire reel, 0.05m, diameter 0.5mm, as drawn, 99.998%
Iron, rod, 500mm, diameter 12.7mm, as drawn, 99.95%
Iron, foil, 50x50mm, thickness 2mm, as rolled, 99.99+%
Iron, tube, 200mm, outside diameter 4.0mm, inside diameter 3.6mm, wall thickness 0.2mm, as drawn, 99.5%
Iron, rod, 200mm, diameter 25mm, as drawn, 98+%
Iron, tube, 100mm, outside diameter 8.0mm, inside diameter 5mm, wall thickness 1.5mm, annealed, 99.5%
Iron, foil, 300x300mm, thickness 1.0mm, as rolled, 99.5%
Iron, foil, 300x300mm, thickness 2.0mm, as rolled, armco« soft ingot 99.8%
Iron, wire reel, 1m, diameter 0.025mm, as drawn, 99.99+%
Iron, foil, 300x300mm, thickness 0.15mm, hard, 99.5%
Iron, foil, 50x50mm, thickness 0.25mm, as rolled, 99.99+%
Iron, wire reel, 1000m, diameter 0.125mm, hard, 99.5%
Iron, foil, 10mm disks, thickness 0.005mm, 99.85%
Iron, tube, 500mm, outside diameter 3.18mm, inside diameter 1.96mm, wall thickness 0.61mm, as drawn, 99.5%
Iron, tube, 200mm, outside diameter 5.0mm, inside diameter 4.5mm, wall thickness 0.25mm, as drawn, 99.5%
Iron, wire reel, 20m, diameter 0.025mm, as drawn, 99.99+%
Iron, wire reel, 1000m, diameter 0.25mm, hard, 99.5%
Iron, wire reel, 250m, diameter 0.25mm, hard, 99.5%