Merck
  • Molybdenum-containing membrane-bound formate dehydrogenase isolated from Citrobacter sp. S-77 having high stability against oxygen, pH, and temperature.
Molybdenum-containing membrane-bound formate dehydrogenase isolated from Citrobacter sp. S-77 having high stability against oxygen, pH, and temperature.
Journal of bioscience and bioengineering (2014-04-23)
Nga T Nguyen, Takeshi Yatabe, Ki-Seok Yoon, Seiji Ogo
ABSTRACT
Membrane-bound formate dehydrogenase (FDH) was purified to homogeneity from a facultative anaerobic bacterium Citrobacter sp. S-77. The FDH from Citrobacter sp. S-77 (FDHS77) was a monomer with molecular mass of approximately 150 kDa. On SDS-PAGE, the purified FDHS77 showed as three different protein bands with molecular mass of approximately 95, 87, and 32 kDa, respectively. Based on the N-terminal amino acid sequence analysis, the sequence alignments observed for the 87 kDa protein band were identical to that of the large subunit of 95 kDa, indicating that the purified FDHS77 consisted of two subunits; a 95 kDa large subunit and a 32 kDa small subunit. The purified FDHS77 in this purification did not contain a heme b subunit, but the FDHS77 showed significant activity for formate oxidation, determined by the Vmax of 30.4 U/mg using benzyl viologen as an electron acceptor. The EPR and ICP-MS spectra indicate that the FDHS77 is a molybdenum-containing enzyme, displaying a remarkable O2-stability along with thermostability and pH resistance. This is the first report of the purification and characterization of a FDH from Citrobacter species.
Sigma-Aldrich
Molybdenum, powder, 10 μm, ≥99.95% trace metals basis
Sigma-Aldrich
Molybdenum, powder, <150 μm, 99.9% trace metals basis
Sigma-Aldrich
Molybdenum, foil, thickness 0.025 mm, ≥99.9% trace metals basis
Sigma-Aldrich
Molybdenum, powder, 1-5 μm, ≥99.9% trace metals basis
Sigma-Aldrich
Molybdenum, nanopowder, <100 nm particle size (TEM), 99.8% trace metals basis
Sigma-Aldrich
Molybdenum, foil, thickness 1.0 mm, ≥99.9% trace metals basis
Sigma-Aldrich
Molybdenum, foil, thickness 0.1 mm, ≥99.9% trace metals basis
Sigma-Aldrich
Molybdenum, powder, <150 μm, 99.99% trace metals basis
Molybdenum, wire reel, 50m, diameter 0.035mm, as drawn, 99.95%
Molybdenum, foil, 300x300mm, thickness 0.30mm, annealed, 99.9%
Molybdenum, foil, 150x150mm, thickness 0.5mm, annealed, 99.9%
Molybdenum, foil, 25x25mm, thickness 0.25mm, annealed, 99.9%
Molybdenum, foil, 0.5m coil, thickness 0.008mm, 99.9%
Molybdenum, wire reel, 50m, diameter 0.8mm, annealed, 99.95%
Molybdenum, foil, 25x25mm, thickness 0.30mm, annealed, 99.9%
Molybdenum, foil, 50x50mm, thickness 2.0mm, annealed, 99.9%
Molybdenum, wire reel, 50m, diameter 0.05mm, annealed, 99.95%
Molybdenum, wire reel, 5m, diameter 1.0mm, annealed, 99.95%
Molybdenum, foil, 50x50mm, thickness 0.125mm, annealed, 99.9%
Molybdenum, wire reel, 500m, diameter 0.025mm, hard, 99.95%
Molybdenum, foil, 25x25mm, thickness 0.5mm, annealed, 99.9%
Molybdenum, foil, 0.2m coil, thickness 0.008mm, 99.9%
Molybdenum, wire reel, 1m, diameter 0.7mm, annealed, 99.95%
Molybdenum, wire reel, 50m, diameter 0.03mm, hard, 99.95%
Molybdenum, foil, 300x300mm, thickness 0.5mm, annealed, 99.9%
Molybdenum, foil, 8mm disks, thickness 0.20mm, annealed, 99.9%
Molybdenum, rod, 100mm, diameter 3.0mm, centerless ground, 99.9%
Molybdenum, foil, 4mm disks, thickness 0.75mm, annealed, 99.9%
Molybdenum, foil, 50mm disks, thickness 0.01mm, 99.9%
Molybdenum, foil, 6mm disks, thickness 0.15mm, annealed, 99.9%