Merck
  • Neoadjuvant therapy combined with a BMP regimen for treating penile cancer patients with lymph node metastasis: a retrospective study in China.
Neoadjuvant therapy combined with a BMP regimen for treating penile cancer patients with lymph node metastasis: a retrospective study in China.
Journal of cancer research and clinical oncology (2014-06-08)
Benkui Zou, Zengchi Han, Zhendan Wang, Jiasheng Bian, Jian Xu, Huansheng Wang, Mingshan Yang, Fajun Pei, Sheng Li, Zhonghua Xu
ABSTRACT
Combination chemotherapy is emerging in the management of advanced penile cancer. However, evidence-based chemotherapeutic regimens in the current guidelines are lacking. The aim of this study was to evaluate the efficacy of preoperative neoadjuvant chemotherapy combined with a BMP regimen including bleomycin (BLM), methopterin (MTX) and cisplatin (DDP) for treating advanced penile cancer patients. We retrospectively audited the clinical and follow-up data of 24 penile cancer patients with fixed inguinal lymph node metastasis that were admitted in our hospital from 2001 to 2010 and received preoperative neoadjuvant chemotherapy. A total of 24 patients with advanced penile cancer (pN3) were recruited in this study. All patients received preoperative neoadjuvant chemotherapy combined with a BMP regimen. The average cycle of chemotherapy was two cycles (range 1-4 cycles). Among 24 adjuvant cases, 15 patients that responded to neoadjuvant chemotherapy underwent penectomy and inguinal lymphadenectomy. In contrast, nine cases did not respond to chemotherapy and received palliative local radiotherapy. Overall, the 1-, 2- and 5-year survival rates were 70.8, 50.0 and 45.8 %, respectively. The 5-year survival rate between the responder and non-responder groups was statistically significant (73.3 vs. 0 %, P < 0.001). Neoadjuvant chemotherapy combined with a BMP regimen followed by surgery is beneficial to patients with advanced penile cancer.
Sigma-Aldrich
cis-Diamineplatinum(II) dichloride, ≥99.9% trace metals basis
Bleomycin sulfate, European Pharmacopoeia (EP) Reference Standard
Methotrexate for system suitability, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Bleomycin sulfate from Streptomyces verticillus, 1.5-2.0 units/mg solid, BioReagent, suitable for cell culture
Sigma-Aldrich
Methotrexate hydrate, powder, BioReagent, suitable for cell culture, ≥98% (HPLC)
Sigma-Aldrich
Methotrexate hydrate, ≥98% (HPLC), powder
Sigma-Aldrich
Bleomycin sulfate from Streptomyces verticillus, crystalline, 1.5-2.0 U/mg
Sigma-Aldrich
Methotrexate, meets USP testing specifications
Sigma-Aldrich
cis-Diammineplatinum(II) dichloride, crystalline
Sigma-Aldrich
Methotrexate hydrate, ≥99.0% (sum of enantiomers, HPLC)
Sigma-Aldrich
Bleomycin sulfate from Streptomyces verticillus, for fluorescence, mixture of bleomycin sulfate salts, lyophilized, powder or crystals, white to off-white
Methotrexate for peak identification, European Pharmacopoeia (EP) Reference Standard
Cisplatin, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Bleomycin sulfate from Streptomyces verticillus, BioXtra, crystalline
Supelco
Methotrexate, Pharmaceutical Secondary Standard; Certified Reference Material
Methotrexate, European Pharmacopoeia (EP) Reference Standard
SAFC
Methotrexate