Merck
  • Corrosion of iron by iodide-oxidizing bacteria isolated from brine in an iodine production facility.
Corrosion of iron by iodide-oxidizing bacteria isolated from brine in an iodine production facility.
Microbial ecology (2014-05-28)
Satoshi Wakai, Kimio Ito, Takao Iino, Yasuyoshi Tomoe, Koji Mori, Shigeaki Harayama
ABSTRACT
Elemental iodine is produced in Japan from underground brine (fossil salt water). Carbon steel pipes in an iodine production facility at Chiba, Japan, for brine conveyance were found to corrode more rapidly than those in other facilities. The corroding activity of iodide-containing brine from the facility was examined by immersing carbon steel coupons in "native" and "filter-sterilized" brine samples. The dissolution of iron from the coupons immersed in native brine was threefold to fourfold higher than that in the filter-sterilized brine. Denaturing gradient gel electrophoresis analyses revealed that iodide-oxidizing bacteria (IOBs) were predominant in the coupon-containing native brine samples. IOBs were also detected in a corrosion deposit on the inner surface of a corroded pipe. These results strongly suggested the involvement of IOBs in the corrosion of the carbon steel pipes. Of the six bacterial strains isolated from a brine sample, four were capable of oxidizing iodide ion (I(-)) into molecular iodine (I(2)), and these strains were further phylogenetically classified into two groups. The iron-corroding activity of each of the isolates from the two groups was examined. Both strains corroded iron in the presence of potassium iodide in a concentration-dependent manner. This is the first report providing direct evidence that IOBs are involved in iron corrosion. Further, possible mechanisms by which IOBs corrode iron are discussed.
Product Description
Iron, IRMM®, certified reference material, 0.5 mm wire
Sigma-Aldrich
Iron, chips, 99.98% trace metals basis
Sigma-Aldrich
Iron, foil, thickness 0.1 mm, ≥99.9% trace metals basis
Sigma-Aldrich
Iron, puriss. p.a., carbonyl-Iron powder, low in magnesium and manganese compounds, ≥99.5% (RT)
Iron, rod, 1000mm, diameter 30mm, as drawn, 98+%
Iron, rod, 150mm, diameter 5.0mm, as drawn, 99.99+%
Iron, foil, 100x100mm, thickness 0.15mm, hard, 99.5%
Iron, rod, 200mm, diameter 8.0mm, as drawn, 99.99+%
Iron, foil, light tested, 50x50mm, thickness 0.025mm, hard, 99.5%
Iron, foil, light tested, 25x25mm, thickness 0.0125mm, 99.99+%
Iron, rod, 100mm, diameter 4.8mm, as drawn, 98+%
Iron, foil, not light tested, 50x50mm, thickness 0.002mm, 99.85%
Iron, rod, 100mm, diameter 100mm, as drawn, armcO« soft ingot 99.8%
Iron, rod, 500mm, diameter 19mm, as drawn, soft ingot 99.8%
Iron, foil, light tested, 25x25mm, thickness 0.075mm, hard, 99.5%
Iron, rod, 1000mm, diameter 12.7mm, as drawn, 98+%
Iron, foil, 50mm disks, thickness 0.38mm, hard, 99.5%
Iron, rod, 500mm, diameter 40mm, as drawn, armcO« soft ingot 99.8%
Iron, foil, not light tested, 150x150mm, thickness 0.009mm, 99.85%
Iron, rod, 100mm, diameter 40mm, as drawn, armcO« soft ingot 99.8%
Iron, foil, not light tested, 50x50mm, thickness 0.01mm, 99.99+%
Iron, rod, 1000mm, diameter 19.0mm, as drawn, 98+%
Iron, foil, 150x150mm, thickness 0.38mm, hard, 99.5%
Iron, insulated wire, 10m, conductor diameter 0.25mm, insulation thickness 0.01mm, polyester insulation
Iron, rod, 500mm, diameter 50mm, as drawn, 98+%
Iron, rod, 200mm, diameter 12.7mm, as drawn, 99.95%
Iron, rod, 200mm, diameter 9.5mm, as drawn, 98+%
Iron, foil, 25x25mm, thickness 1.83mm, Quarter hard, 99.5%
Iron, foil, not light tested, 150x150mm, thickness 0.004mm, 99.85%
Iron, rod, 50mm, diameter 12.7mm, as drawn, 99.95%