Merck
mTOR-Controlled Autophagy Requires Intracellular Ca(2+) Signaling.
PloS one (2013-04-09)
Jean-Paul Decuypere, Dimphny Kindt, Tomas Luyten, Kirsten Welkenhuyzen, Ludwig Missiaen, Humbert De Smedt, Geert Bultynck, Jan B Parys
ABSTRACT
Autophagy is a lysosomal degradation pathway important for cellular homeostasis and survival. Inhibition of the mammalian target of rapamycin (mTOR) is the best known trigger for autophagy stimulation. In addition, intracellular Ca(2+) regulates autophagy, but its exact role remains ambiguous. Here, we report that the mTOR inhibitor rapamycin, while enhancing autophagy, also remodeled the intracellular Ca(2+)-signaling machinery. These alterations include a) an increase in the endoplasmic-reticulum (ER) Ca(2+)-store content, b) a decrease in the ER Ca(2+)-leak rate, and c) an increased Ca(2+) release through the inositol 1,4,5-trisphosphate receptors (IP3Rs), the main ER-resident Ca(2+)-release channels. Importantly, buffering cytosolic Ca(2+) with BAPTA impeded rapamycin-induced autophagy. These results reveal intracellular Ca(2+) signaling as a crucial component in the canonical mTOR-dependent autophagy pathway.