Merck
  • N-Acetyl-2-hydroxy-N'-[methoxy(1-methylindol-2-yl)methyl]benzohydrazide.
N-Acetyl-2-hydroxy-N'-[methoxy(1-methylindol-2-yl)methyl]benzohydrazide.
Acta crystallographica. Section E, Structure reports online (2008-01-01)
Wagee A Yehye, Noorsaadah Abdul Rahman, Azhar Ariffin, Seik Weng Ng
ABSTRACT
In the crystal structure of the title Schiff-base, C(20)H(21)N(3)O(4), the amino group forms an N-H⋯O hydrogen bond to the acetyl group of an adjacent mol-ecule, forming a zigzag chain. The 2-hydr-oxy group is inter-nally hydrogen bonded to the amido group though an O-H⋯O hydrogen bond.
Sigma-Aldrich
1-Methylindole-3-carboxaldehyde, 97%