Merck
  • Synthetic ion transporters can induce apoptosis by facilitating chloride anion transport into cells.
Synthetic ion transporters can induce apoptosis by facilitating chloride anion transport into cells.
Nature chemistry (2014-09-23)
Sung-Kyun Ko, Sung Kuk Kim, Andrew Share, Vincent M Lynch, Jinhong Park, Wan Namkung, Wim Van Rossom, Nathalie Busschaert, Philip A Gale, Jonathan L Sessler, Injae Shin
ABSTRACT
Anion transporters based on small molecules have received attention as therapeutic agents because of their potential to disrupt cellular ion homeostasis. However, a direct correlation between a change in cellular chloride anion concentration and cytotoxicity has not been established for synthetic ion carriers. Here we show that two pyridine diamide-strapped calix[4]pyrroles induce coupled chloride anion and sodium cation transport in both liposomal models and cells, and promote cell death by increasing intracellular chloride and sodium ion concentrations. Removing either ion from the extracellular media or blocking natural sodium channels with amiloride prevents this effect. Cell experiments show that the ion transporters induce the sodium chloride influx, which leads to an increased concentration of reactive oxygen species, release of cytochrome c from the mitochondria and apoptosis via caspase activation. However, they do not activate the caspase-independent apoptotic pathway associated with the apoptosis-inducing factor. Ion transporters, therefore, represent an attractive approach for regulating cellular processes that are normally controlled tightly by homeostasis.
Sigma-Aldrich
N-Acetyl-Asp-Glu-Val-Asp-al, ≥95%, powder
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Hydrochloric acid, BioReagent, 36.5-38.0%
Sigma-Aldrich
Caspase 3 Assay Kit, Colorimetric
Sigma-Aldrich
Thiourea, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Potassium chloride, BioXtra, ≥99.0%
Sigma-Aldrich
Amiloride hydrochloride hydrate, ≥98% (HPLC), powder
Sigma-Aldrich
DCFHDA, BioReagent, suitable for fluorescence, ≥95% (HPLC)
Sigma-Aldrich
Chloroform, anhydrous, ≥99%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Dimethyl sulfoxide-d6, "100%", 99.96 atom % D
Sigma-Aldrich
Pyrrole, reagent grade, 98%
Sigma-Aldrich
Caspase 3 Substrate
Sigma-Aldrich
2-Oleoyl-1-palmitoyl-sn-glycero-3-phosphocholine, ≥99.0% (TLC)
Sigma-Aldrich
Acetone, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%
Sigma-Aldrich
2,6-Pyridinedicarboxamide, 97%
Sigma-Aldrich
1-(Ethoxycarbonylmethyl)-6-methoxyquinolinium bromide, suitable for fluorescence, ≥97.0% (TLC)
Sigma-Aldrich
Tetraethylammonium chloride, BioUltra, ≥99.0% (AT)
Sigma-Aldrich
4-Nitroaniline, ≥99%
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
Urea, BioXtra, pH 7.5-9.5 (20 °C, 5 M in H2O)
Sigma-Aldrich
Dimethyl sulfoxide
Sigma-Aldrich
JC-1, powder or solid (Crystals)
Sigma-Aldrich
Potassium sulfate, BioXtra, ≥99.0%
Sigma-Aldrich
N,N′-Dimethyl-9,9′-biacridinium dinitrate, used as chemiluminescent reagent
Sigma-Aldrich
Caspase 3 Assay Kit, Fluorimetric
Sigma-Aldrich
N-Acetyl-Asp-Glu-Val-Asp p-nitroanilide, ≥97% (HPLC), powder
Sigma-Aldrich
Prodigiosin hydrochloride, from Serratia marcescens, ≥98% (HPLC), powder
Sigma-Aldrich
Cesium chloride, ReagentPlus®, 99.9%
Sigma-Aldrich
Pyridine, biotech. grade, ≥99.9%