Merck
  • Rapid screening of oleuropein from olive leaves using matrix solid-phase dispersion and high-performance liquid chromatography.
Rapid screening of oleuropein from olive leaves using matrix solid-phase dispersion and high-performance liquid chromatography.
Journal of AOAC International (2014-08-26)
Marzieh Rashidipour, Rouhollah Heydari, Alireza Feizbakhsh, Payman Hashemi
ABSTRACT
Matrix solid-phase dispersion (MSPD) methodology as a quick and easy extraction method has been developed to extract oleuropein from Olea europaea leaves. This method has been compared with conventional maceration and Soxhlet extraction (SOXE) methods. Variables affecting MSPD, such as the sorbent material, the ratio of sample to sorbent material, elution solvent, and volume of the elution solvent, have been fully evaluated and optimized. Oleuropein was successfully extracted by silica sorbent with 350 microL of dichloromethane-methanol (70 + 30, v/v) as the elution solvent. The calibration curve showed good linearity (r2 = 0.9985) and precision (RSD <4.5%) in the concentration range of 0.5-100 microg/mL for oleuropein. The LOD and LOQ were 0.14 and 0.5 microg/mL, respectively. The recoveries were in the range of 88.0-94.0% with RSD values ranging from 6.0 to 9.8%. The extraction yields obtained by the MSPD method were comparable with those obtained by the maceration and SOXE methods. The reported MSPD extraction method was revealed to be eco-friendly, simpler, and faster than conventional methods used to quantify oleuropein in O. europaea leaves.
Product Description
Supelco
Methanol, analytical standard
Supelco
1,4-Dioxane, analytical standard
Supelco
Acetone, analytical standard
Sigma-Aldrich
Phosphoric acid solution, 85 wt. % in H2O, FCC, FG
Sigma-Aldrich
1,4-Dioxane, anhydrous, 99.8%, contains <=25 ppm BHT as stabilizer
Sigma-Aldrich
Diethyl ether, contains 1 ppm BHT as inhibitor, anhydrous, ≥99.7%
Sigma-Aldrich
Diethyl ether
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Acetone, natural, ≥97%
Sigma-Aldrich
Acetone, ≥99%, meets FCC analytical specifications
Supelco
Diethyl ether, analytical standard
Sigma-Aldrich
Methanol
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.9%
Supelco
Acetone, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Phosphoric acid solution, 85% in D2O (99.9 atom % D), NMR tube size 4.2 mm × 8 in. , WGS-5BL Coaxial NMR tube
Supelco
Ethanol solution, certified reference material, 2000 μg/mL in methanol
Supelco
Dehydrated Alcohol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Acetone, histological grade, ≥99.5%
Sigma-Aldrich
Acetone, ACS reagent, ≥99.5%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Acetone, Laboratory Reagent, ≥99.5%
Sigma-Aldrich
Acetone, ACS reagent, ≥99.5%
Sigma-Aldrich
1,4-Dioxane, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.5% (GC)
Sigma-Aldrich
Acetone, puriss., meets analytical specification of Ph. Eur., BP, NF, ≥99% (GC)
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Diethyl ether, reagent grade, ≥98%, contains ≤2% ethanol and ≤10ppm BHT as inhibitor
Sigma-Aldrich
Diethyl ether, contains BHT as inhibitor, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
Methanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
Methanol, puriss., meets analytical specification of Ph Eur, ≥99.7% (GC)
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%