Merck
  • Multiple nutrient stresses at intersecting Pacific Ocean biomes detected by protein biomarkers.
Multiple nutrient stresses at intersecting Pacific Ocean biomes detected by protein biomarkers.
Science (New York, N.Y.) (2014-09-06)
Mak A Saito, Matthew R McIlvin, Dawn M Moran, Tyler J Goepfert, Giacomo R DiTullio, Anton F Post, Carl H Lamborg
ABSTRACT
Marine primary productivity is strongly influenced by the scarcity of required nutrients, yet our understanding of these nutrient limitations is informed by experimental observations with sparse geographical coverage and methodological limitations. We developed a quantitative proteomic method to directly assess nutrient stress in high-light ecotypes of the abundant cyanobacterium Prochlorococcus across a meridional transect in the central Pacific Ocean. Multiple peptide biomarkers detected widespread and overlapping regions of nutritional stress for nitrogen and phosphorus in the North Pacific Subtropical Gyre and iron in the equatorial Pacific. Quantitative protein analyses demonstrated simultaneous stress for these nutrients at biome interfaces. This application of proteomic biomarkers to diagnose ocean metabolism demonstrated Prochlorococcus actively and simultaneously deploying multiple biochemical strategies for low-nutrient conditions in the oceans.
Sigma-Aldrich
Cobalt, Carbon coated magnetic, nanopowder, <50 nm particle size (TEM), ≥99%
Sigma-Aldrich
Iron, puriss. p.a., carbonyl-Iron powder, low in magnesium and manganese compounds, ≥99.5% (RT)
Iron, IRMM®, certified reference material, 0.5 mm wire
Sigma-Aldrich
Iron, foil, thickness 0.1 mm, ≥99.9% trace metals basis
Sigma-Aldrich
Cobalt, foil, thickness 0.1 mm, ≥99.99%
Sigma-Aldrich
Cobalt, granular, 99.99% trace metals basis
Sigma-Aldrich
Cobalt, wire, diam. 1.0 mm, 99.995% trace metals basis
Sigma-Aldrich
Cobalt, foil, thickness 0.1 mm, 99.95% trace metals basis
Sigma-Aldrich
Cobalt, powder, <150 μm, ≥99.9% trace metals basis
Sigma-Aldrich
Iron, chips, 99.98% trace metals basis
Sigma-Aldrich
Cobalt, foil, thickness 1.0 mm, 99.95% trace metals basis
Sigma-Aldrich
Cobalt, rod, diam. 5.0 mm, 99.95% trace metals basis
Sigma-Aldrich
Cobalt, pieces, 99.5% trace metals basis
Sigma-Aldrich
Cobalt, powder, 2 μm particle size, 99.8% trace metals basis
Iron, wire reel, 5m, diameter 0.025mm, as drawn, 99.99+%
Iron, wire reel, 1m, diameter 0.5mm, as drawn, 99.998%
Iron, wire reel, 0.5m, diameter 0.25mm, hard, 99.99+%
Iron, wire reel, 0.5m, diameter 1.0mm, as drawn, 99.99+%
Iron, wire reel, 5m, diameter 0.25mm, hard, 99.99+%
Iron, wire reel, 50m, diameter 0.5mm, as drawn, 99.95%
Iron, wire reel, 2m, diameter 0.25mm, annealed, 99.99+%
Iron, wire reel, 0.05m, diameter 0.5mm, as drawn, 99.998%
Iron, rod, 500mm, diameter 12.7mm, as drawn, 99.95%
Iron, foil, 50x50mm, thickness 2mm, as rolled, 99.99+%
Iron, tube, 200mm, outside diameter 4.0mm, inside diameter 3.6mm, wall thickness 0.2mm, as drawn, 99.5%
Iron, rod, 200mm, diameter 25mm, as drawn, 98+%
Iron, tube, 100mm, outside diameter 8.0mm, inside diameter 5mm, wall thickness 1.5mm, annealed, 99.5%
Iron, foil, 300x300mm, thickness 1.0mm, as rolled, 99.5%
Iron, foil, 300x300mm, thickness 2.0mm, as rolled, armco« soft ingot 99.8%
Cobalt, foil, 15mm disks, thickness 0.003mm, 99.9%