Merck
  • A comparative assessment of small-head metal-on-metal and ceramic-on-polyethylene total hip replacement.
A comparative assessment of small-head metal-on-metal and ceramic-on-polyethylene total hip replacement.
The bone & joint journal (2014-07-06)
A Lübbeke, A Gonzalez, G Garavaglia, C Roussos, A Bonvin, R Stern, R Peter, P Hoffmeyer
ABSTRACT
Large-head metal-on-metal (MoM) total hip replacements (THR) have given rise to concern. Comparative studies of small-head MoM THRs over a longer follow-up period are lacking. Our objective was to compare the incidence of complications such as infection, dislocation, revision, adverse local tissue reactions, mortality and radiological and clinical outcomes in small-head (28 mm) MoM and ceramic-on-polyethylene (CoP) THRs up to 12 years post-operatively. A prospective cohort study included 3341 THRs in 2714 patients. The mean age was 69.1 years (range 24 to 98) and 1848 (55.3%) were performed in women, with a mean follow-up of 115 months (18 to 201). There were 883 MoM and 2458 CoP bearings. Crude incidence rates (cases/1000 person-years) were: infection 1.3 vs 0.8; dislocation 3.3 vs 3.1 and all-cause revision 4.3 vs 2.2, respectively. There was a significantly higher revision rate after ten years (adjusted hazard ratio 9.4; 95% CI 2.6 to 33.6) in the MoM group, and ten of 26 patients presented with an adverse local tissue reaction at revision. No differences in mortality, osteolysis or clinical outcome were seen. In conclusion, we found similar results for small-head MoM and CoP bearings up to ten years post-operatively, but after ten years MoM THRs had a higher risk of all-cause revision. Furthermore, the presence of an adverse response to metal debris seen in the small-head MOM group at revision is a cause for concern.
Product Description
Supelco
Polyethylene, analytical standard, for GPC, 2,000
Sigma-Aldrich
Polyethylene, average Mw ~4,000 by GPC, average Mn ~1,700 by GPC
Sigma-Aldrich
Polyethylene, Medium density
Sigma-Aldrich
Polyethylene, Ultra-high molecular weight, average Mw 3,000,000-6,000,000
Sigma-Aldrich
Polyethylene, Ultra-high molecular weight, surface-modified, powder, 125 μm avg. part. size
Sigma-Aldrich
Polyethylene, Ultra-high molecular weight, surface-modified, powder, 34-50 μm particle size
Sigma-Aldrich
Polyethylene, low density, melt index 25 g/10 min (190°C/2.16kg)
Sigma-Aldrich
Polyethylene, Linear low density, melt index 1.0 g/10 min (190°C/2.16kg)
Sigma-Aldrich
Polyethylene, High density, melt index 12 g/10 min (190 °C/2.16kg)
Sigma-Aldrich
Polyethylene, High density, melt index 2.2 g/10 min (190 °C/2.16kg)
Polyethylene (LDPE), ERM®, certified reference material