Merck
  • Hollow iron-silica nanoshells for enhanced high intensity focused ultrasound.
Hollow iron-silica nanoshells for enhanced high intensity focused ultrasound.
The Journal of surgical research (2014-06-29)
Alexander Liberman, Zhe Wu, Christopher V Barback, Robert D Viveros, James Wang, Lesley G Ellies, Robert F Mattrey, William C Trogler, Andrew C Kummel, Sarah L Blair
ABSTRACT
High intensity-focused ultrasound (HIFU) is an alterative ablative technique currently being investigated for local treatment of breast cancer and fibroadenomas. Current HIFU therapies require concurrent magnetic resonance imaging monitoring. Biodegradable 500 nm perfluoropentane-filled iron-silica nanoshells have been synthesized as a sensitizing agent for HIFU therapies, which aid both mechanical and thermal ablation of tissues. In low duty cycle high-intensity applications, rapid tissue damage occurs from mechanical rather than thermal effects, which can be monitored closely by ultrasound obviating the need for concurrent magnetic resonance imaging. Iron-silica nanoshells were synthesized by a sol-gel method on polystyrene templates and calcined to yield hollow nanoshells. The nanoshells were filled with perfluoropentane and injected directly into excised human breast tumor, and intravenously (IV) into healthy rabbits and Py8119 tumor-bearing athymic nude mice. HIFU was applied at 1.1 MHz and 3.5 MPa at a 2% duty cycle to achieve mechanical ablation. Ex vivo in excised rabbit livers, the time to visually observable damage with HIFU was 20 s without nanoshells and only 2 s with nanoshells administered IV before sacrifice. Nanoshells administered IV into nude mice with xenograft tumors were activated in vivo by HIFU 24 h after administration. In this xenograft model, applied HIFU resulted in a 13.6 ± 6.1 mm(3) bubble cloud with the IV injected particles and no bubble cloud without particles. Iron-silica nanoshells can reduce the power and time to perform HIFU ablative therapy and can be monitored by ultrasound during low duty cycle operation.
Sigma-Aldrich
Silica, nanopowder, 99.8% trace metals basis
Sigma-Aldrich
Silica, mesostructured, MCM-41 type (hexagonal)
Sigma-Aldrich
Silica, nanoparticles, mesoporous, 200 nm particle size, pore size 4 nm
Sigma-Aldrich
Silicon dioxide, nanopowder (spherical, porous), 5-20 nm particle size (TEM), 99.5% trace metals basis
Sigma-Aldrich
Silicon dioxide, nanopowder, 10-20 nm particle size (BET), 99.5% trace metals basis
Sigma-Aldrich
Iron, puriss. p.a., carbonyl-Iron powder, low in magnesium and manganese compounds, ≥99.5% (RT)
Sigma-Aldrich
LUDOX® TM-40 colloidal silica, 40 wt. % suspension in H2O
Sigma-Aldrich
LUDOX® SM colloidal silica, 30 wt. % suspension in H2O
Sigma-Aldrich
LUDOX® HS-30 colloidal silica, 30 wt. % suspension in H2O
Sigma-Aldrich
LUDOX® LS colloidal silica, 30 wt. % suspension in H2O
Iron, IRMM®, certified reference material, 0.5 mm wire
Sigma-Aldrich
Iron, foil, thickness 0.1 mm, ≥99.9% trace metals basis
Sigma-Aldrich
LUDOX® CL colloidal silica, 30 wt. % suspension in H2O
Sigma-Aldrich
Silica
Sigma-Aldrich
Iron, chips, 99.98% trace metals basis
Sigma-Aldrich
Silica, mesostructured, MSU-F (cellular foam)
Iron, foil, light tested, 25x25mm, thickness 0.05mm, hard, 99.5%
Iron, foil, 6mm disks, thickness 0.5mm, hard, 99.5%
Iron, foil, 50mm disks, thickness 0.25mm, hard, 99.5%
Iron, foil, light tested, 100x100mm, thickness 0.025mm, as rolled, 99.99+%
Iron, foil, 8mm disks, thickness 0.05mm, as rolled, 99.99+%
Iron, foil, not light tested, 100x100mm, thickness 0.003mm, 99.85%
Iron, foil, 6mm disks, thickness 0.125mm, hard, 99.5%
Iron, foil, not light tested, 100x100mm, thickness 0.005mm, 99.85%
Iron, foil, 50mm disks, thickness 0.38mm, hard, 99.5%
Iron, foil, light tested, 25x25mm, thickness 0.075mm, hard, 99.5%
Iron, foil, 6mm disks, thickness 0.020mm, 99.85%
Iron, foil, light tested, 100x100mm, thickness 0.025mm, hard, 99.5%
Iron, foil, 6mm disks, thickness 0.125mm, as rolled, 99.99+%
Iron, foil, 8mm disks, thickness 0.05mm, hard, 99.5%