Merck
  • Optimization for the removal of orthophosphate from aqueous solution by chemical precipitation using ferrous chloride.
Optimization for the removal of orthophosphate from aqueous solution by chemical precipitation using ferrous chloride.
Environmental technology (2014-06-25)
Ju-Suk An, Ye-Ji Back, Ki-Chul Kim, Ran Cha, Tae-Young Jeong, Hyung-Keun Chung
ABSTRACT
The precipitation reaction between the orthophosphate and Fe2+ ions was studied to describe the optimum condition for the removal of orthophosphate from the aqueous solution. The effects of pH, Fe:P molar ratio, and alkalinity were evaluated for the initial orthophosphate concentrations in the range from 1.55 to 31.00 mg/L - PO4(3-) -P. The optimum pH was found to be 8.0 in all orthophosphate concentration ranges. When the stoichiometric moles of Fe2+ were added, the removal efficiencies were significantly less than the theoretical values. It is likely that the precipitation of Fe(OH)2(s) is partially formed. For the initial orthophosphate concentration of 3.10 mg/L PO4(3-) -P or greater, the removal efficiencies with the Fe:P molar ratio of 3.0:1.0 approached to the theoretical values, yielding greater than 98.5%. If the molar ratio of Fe:P was great enough, the precipitation reaction was completed within 1 h. As the alkalinity increases, the experimental removal efficiencies were significantly greater than the theoretical values. This is because the formation of vivianite is favoured over FeCO3(s). Finally, it was demonstrated that the orthophosphate (1.40-6.80 mg/L PO4(3-) -P) in the secondary effluents from wastewater treatment plants was effectively removed by dosing sufficient amount of Fe2+ ions.
Sigma-Aldrich
Iron(II) chloride, 98%
Sigma-Aldrich
Iron(II) chloride, AnhydroBeads, −10 mesh, 99.9% trace metals basis
Sigma-Aldrich
Iron(II) chloride tetrahydrate, 99.99% trace metals basis
Sigma-Aldrich
Iron(II) chloride, AnhydroBeads, −10 mesh, 99.998% trace metals basis
Sigma-Aldrich
Iron(II) chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
Iron(II) chloride tetrahydrate, puriss. p.a., ≥99.0% (RT)
Sigma-Aldrich
Iron(II) chloride tetrahydrate, ReagentPlus®, 98%