Merck
  • Fluoride exposure regulates the elongation phase of protein synthesis in cultured Bergmann glia cells.
Fluoride exposure regulates the elongation phase of protein synthesis in cultured Bergmann glia cells.
Toxicology letters (2014-06-24)
Marco Flores-Méndez, Diana Ramírez, Nely Alamillo, Luisa C Hernández-Kelly, Luz María Del Razo, Arturo Ortega
ABSTRACT
Fluoride is an environmental pollutant present in dental products, food, pesticides and water. The latter, is the greatest source of exposure to this contaminant. Structural and functional damages to the central nervous system are present in exposed population. An established consequence of the neuronal is the release of a substantial amount of glutamate to the extracellular space, leading to an excitotoxic insult. Glutamate exerts its actions through the activation of specific plasma membrane receptors and transporters present in neurons and in glia cells and it is the over-activation of glutamate receptors and transporters, the biochemical hallmark of neuronal and oligodendrocyte cell death. In this context, taking into consideration that fluoride leads to degeneration of cerebellar cells, we took the advantage of the well-established model of cerebellar Bergmann glia cultures to gain insight into the molecular mechanisms inherent to fluoride neurotoxicity that might be triggered in glia cells. We could establish that fluoride decreases [(35)S]-methionine incorporation into newly synthesized polypeptides, in a time-dependent manner, and that this halt in protein synthesis is the result of a decrease in the elongation phase of translation, mediated by an augmentation of eukaryotic elongation factor 2 phosphorylation. These results favor the notion of glial cells as targets of fluoride toxicity and strengthen the idea of a critical involvement of glia cells in the function and dysfunction of the brain.
Product Description
Supelco
L-Methionine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
L-Glutamine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
L-Glutamine
Supelco
L-Glutamine, Pharmaceutical Secondary Standard; Certified Reference Material
SAFC
L-Methionine
Sigma-Aldrich
L-Methionine, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
Sodium fluoride, anhydrous, powder, 99.99% trace metals basis
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.9% trace metals basis
Supelco
Fluoride ion solution for ISE, 0.1 M F-, analytical standard (for ion-selective electrodes)
SAFC
L-Glutamine
Sigma-Aldrich
DL-Methionine, 99%, FCC, FG
Sigma-Aldrich
Sodium fluoride, 99.99% trace metals basis
Sigma-Aldrich
Magnesium chloride, powder, <200 μm
Sigma-Aldrich
L-Glutamine, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
Sigma-Aldrich
Magnesium chloride, BioReagent, suitable for insect cell culture, ≥97.0%
Sigma-Aldrich
Magnesium chloride, anhydrous, ≥98%
Sigma-Aldrich
L-Glutamine, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium fluoride, BioXtra, ≥99%
Sigma-Aldrich
L-Methionine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 99.0-101.0%
Sigma-Aldrich
Sodium fluoride, BioReagent, suitable for insect cell culture, ≥99%
Sigma-Aldrich
L-Aspartic acid potassium salt, ≥98% (HPLC)
Sigma-Aldrich
DL-Methionine, ≥99%
Sigma-Aldrich
DL-Methionine, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99%
Sigma-Aldrich
L-Methionine, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
L-Glutamine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
Sodium fluoride 0.5 M solution
DL-Methionine, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
DL-Methionine, ≥99.0% (NT)
Sigma-Aldrich
L-Glutamine, BioUltra, ≥99.5% (NT)