Merck
  • Oral doses of α-retinyl ester track chylomicron uptake and distribution of vitamin A in a male piglet model for newborn infants.
Oral doses of α-retinyl ester track chylomicron uptake and distribution of vitamin A in a male piglet model for newborn infants.
The Journal of nutrition (2014-06-20)
Napaporn Riabroy, Sherry A Tanumihardjo
ABSTRACT
α-Retinol has utility in determining chylomicron trafficking of vitamin A to tissues given that it will not be recirculated in blood on retinol binding protein (RBP). In this study, α-retinol was used as a chylomicron tag to investigate short-term uptake from high-dose supplements given to piglets as a model for neonates. The distribution of orally administered α-retinol doses in liver and extrahepatic tissues was assessed at varying times after dosing. Male piglets (n = 24 per group) from vitamin A-depleted sows were orally given 26.2 or 52.4 μmol of α-retinyl acetate, the molar equivalent of 25,000 and 50,000 IU of vitamin A, respectively. Tissues were collected and analyzed by HPLC. Lung (6.46 ± 2.94 nmol/g), spleen (22.1 ± 11.3 nmol/g), and adrenal gland (17.0 ± 11.2 nmol/g) α-retinol concentrations peaked at 7 h after dosing, and, by 7 d, α-retinol was essentially cleared from these tissues (≤0.25 ± 0.12 nmol/g). This demonstrates that the lung, spleen, and adrenal gland receive substantial vitamin A from chylomicra to maintain concentrations. Conversely, storage of α-retinol in the liver reached a plateau at 24 h (1.72 ± 0.58 μmol/liver) and was retained through 7 d (2.10 ± 0.38 μmol/liver) (P > 0.05). This indicates that α-retinol was not substantially utilized locally in the liver nor transported out from the liver via RBP. In serum, the majority of α-retinol was in the ester form, which confirms that α-retinol does not bind to RBP but does circulate. α-Retinyl esters were detectable at 7 d in the serum but were not different from baseline. Collectively, these data suggest that crucial immune organs need constant dietary intake to maintain vitamin A concentrations because α-retinol was quickly taken up by tissues and decreased to baseline in all tissues except long-term storage in the liver.
Sigma-Aldrich
Retinol, BioXtra, ≥97.5% (HPLC), ~3100 U/mg
Sigma-Aldrich
Retinol, synthetic, ≥95% (HPLC), (Powder or Powder with Lumps)
Sigma-Aldrich
Retinol, ≥95.0% (HPLC), ~2700 U/mg
Supelco
Retinyl Acetate (Vitamin A Acetate), Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Retinyl acetate, analytical standard
Sigma-Aldrich
Retinyl acetate, synthetic, matrix dispersion, 475,000-650,000 USP units/g
Sigma-Aldrich
Retinyl acetate, synthetic, crystalline solid or supercooled liquid
Sigma-Aldrich
Retinyl acetate, solid or viscous liquid, BioReagent, synthetic, suitable for cell culture
Retinol acetate, European Pharmacopoeia (EP) Reference Standard
USP
Retinyl acetate, United States Pharmacopeia (USP) Reference Standard, solution of retinyl acetate (vitamin A) in peanut oil