Merck
  • β-cyclodextrin and curcumin, a potent cocktail for disaggregating and/or inhibiting amyloids: a case study with α-synuclein.
β-cyclodextrin and curcumin, a potent cocktail for disaggregating and/or inhibiting amyloids: a case study with α-synuclein.
Biochemistry (2014-06-17)
Saurabh Gautam, Sandip Karmakar, Abhik Bose, Pramit K Chowdhury
ABSTRACT
Aggregation of α-synuclein has been implicated in Parkinson's disease (PD). While many compounds are known to inhibit α-synuclein aggregation, dissolution of aggregates into their constituent monomers cannot be readily achieved. In this study, using a range of techniques, we have shown that an optimized cocktail of curcumin and β-cyclodextrin, at appreciably low concentrations, not only inhibited aggregation but also broke up the preformed aggregates almost completely. We propose that these compounds exhibit synergy in their action and thus provide us with the exciting prospect of working toward the development of a suitable drug candidate for prevention and treatment of PD.
Sigma-Aldrich
β-Cyclodextrin, produced by Wacker Chemie AG, Burghausen, Germany, ≥95.0% (HPLC)
USP
Beta Cyclodextrin, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
β-Cyclodextrin, produced by Wacker Chemie AG, Burghausen, Germany
Sigma-Aldrich
β-Cyclodextrin, powder, BioReagent, suitable for cell culture, ≥97%
Sigma-Aldrich
β-Cyclodextrin, ≥97%
Sigma-Aldrich
Thioflavine S, practical grade
Sigma-Aldrich
β-Cyclodextrin, Produced by Wacker Chemie AG, Burghausen, Germany, Life ScienceProduced by Wacker Chemie AG, Burghausen, Germany, Life Science, ≥98.0% (HPLC)
Sigma-Aldrich
Thioflavin T, used as stain for amyloid
Supelco
Beta-Cyclodextrin, Pharmaceutical Secondary Standard; Certified Reference Material
Betadex, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Curcumin, ≥94% (curcuminoid content), ≥80% (Curcumin)
Supelco
Curcumin, analytical standard
Sigma-Aldrich
Curcumin, from Curcuma longa (Turmeric), powder
USP
Curcumin, United States Pharmacopeia (USP) Reference Standard
Supelco
Curcumin, matrix substance for MALDI-MS, ≥99.5% (HPLC)
Curcumin, primary reference standard