Merck
  • Left ventricular mass changes after renal transplantation: influence of dietary sodium and change in serum uric acid.
Left ventricular mass changes after renal transplantation: influence of dietary sodium and change in serum uric acid.
Transplantation (2014-06-14)
Guilhem du Cailar, Carole Oudot, Pierre Fesler, Albert Mimran, Benjamin Bonnet, Vincent Pernin, Jean Ribstein, Georges Mourad
ABSTRACT
We hypothesized that dietary sodium may modulate the effect of systolic blood pressure and other nonhemodynamic factors, such as high uric acid and renal dysfunction, on changes in the left ventricular mass after renal transplantation. The objective of the present 3-year follow-up longitudinal study was to assess the concomitant influence of these factors on changes in the left ventricular mass after renal transplantation. Twenty-four-hour urinary sodium excretion, glomerular filtration rate (isotopic clearance), and left ventricular mass (echocardiography) assessment were done in 165 renal transplant patients during the first year and after a follow-up of 3 years after renal transplantation. At follow-up, therapy of hypertension was associated with normalization of blood pressure in 64% and a decrease in the prevalence of left ventricular hypertrophy from 66% to 56%. At baseline and follow-up, systolic blood pressure, sodium intake, and serum uric acid emerged as independent and significant determinants of the final left ventricular mass index. When the population was divided according to sex-specific tertiles of the final 24-hr urinary sodium excretion, the relationship between change in serum uric acid during follow-up, final left ventricular mass index, and final glomerular filtration rate was significant only on the highest tertile of 24-hr urinary sodium excretion. The decrease in the prevalence of left ventricular hypertrophy after renal transplantation is blunted by high sodium intake. Persistence of the left ventricular hypertrophy may result from the combined adverse influences of excessive dietary sodium intake and increased serum uric acid during follow-up despite pharmacological control of blood pressure.
Sigma-Aldrich
Uric acid, BioXtra, ≥99% (HPLC)
Sigma-Aldrich
Uric acid, ≥99%, crystalline