Merck
  • Conventional and novel peripheral blood iron markers compared against bone marrow in Malawian children.
Conventional and novel peripheral blood iron markers compared against bone marrow in Malawian children.
Journal of clinical pathology (2014-06-12)
Femkje A M Jonker, Michael Boele van Hensbroek, Tjalling Leenstra, Raymond J W M Vet, Bernard J Brabin, Nelson Maseko, Montfort B Gushu, Mercy Emana, Rob Kraaijenhagen, Harold Tjalsma, Dorine W Swinkels, Job C J Calis
ABSTRACT
Iron deficiency is an important child health problem. Its diagnosis in areas of high infection exposure remains complicated as inflammation may interfere with the accuracy of peripheral iron markers. With this study, we aimed to validate the conventional iron markers and two novel iron markers, hepcidin and Red blood cell Size Factor (RSf), against the reference standard of iron status, bone marrow iron, in children living in an infectious setting. We compared ferritin, soluble transferrin receptor, Soluble Transferrin Log-Ferritin Index (sTfR-F), mean cellular volume, mean cellular haemoglobin concentration, hepcidin and RSf, against bone marrow iron in 87 healthy Malawian children (6-66 months) scheduled for elective surgery. Of all children, 44.8% had depleted bone marrow iron stores. Using optimised cut-offs, ferritin (<18 µg/L) and sTfR-F (>1.85) best predicted depleted iron stores with a sensitivity/specificity of 73.7%/77.1% and 72.5%/75.0%, respectively. Hepcidin (<1.4 nmol/L) was a moderate sensitive marker (73.0%) although specificity was 54.2%; RSf poorly predicted depleted iron stores. We provide the first bone marrow-validated data on peripheral iron markers in African children, and showed ferritin and sTfR-F best predicted iron status. Using appropriately defined cut-offs, these indicators can be applied in surveillance and research. As their accuracy is limited for clinical purposes, more reliable iron biomarkers are still required in African children.
Sigma-Aldrich
Iron, chips, 99.98% trace metals basis
Sigma-Aldrich
Iron, foil, thickness 0.1 mm, ≥99.9% trace metals basis
Sigma-Aldrich
Ferritin from equine spleen, Type I, saline solution
Iron, IRMM®, certified reference material, 0.5 mm wire
Sigma-Aldrich
Iron, puriss. p.a., carbonyl-Iron powder, low in magnesium and manganese compounds, ≥99.5% (RT)
Iron, foil, 25mm disks, thickness 0.015mm, 99.85%
Iron, foil, 15mm disks, thickness 0.1mm, hard, 99.5%
Iron, foil, 25mm disks, thickness 0.01mm, 99.99+%
Iron, foil, 20m coil, thickness 0.038mm, hard, 99.5%
Iron, foil, 25mm disks, thickness 0.05mm, hard, 99.5%
Iron, foil, 10mm disks, thickness 0.38mm, hard, 99.5%
Iron, foil, 25mm disks, thickness 0.05mm, as rolled, 99.99+%
Iron, foil, 50mm disks, thickness 0.038mm, hard, 99.5%
Iron, foil, 10mm disks, thickness 0.9mm, as rolled, 99.5%
Iron, foil, 25mm disks, thickness 0.125mm, hard, 99.5%
Iron, foil, 1m coil, thickness 0.05mm, hard, 99.5%
Iron, foil, 1m coil, thickness 0.38mm, hard, 99.5%
Iron, foil, 4mm disks, thickness 0.025mm, as rolled, 99.99+%
Iron, foil, 50mm disks, thickness 0.075mm, hard, 99.5%
Iron, foil, 4mm disks, thickness 0.20mm, hard, 99.5%
Iron, foil, 4mm disks, thickness 0.5mm, hard, 99.5%
Iron, wire reel, 0.05m, diameter 0.5mm, as drawn, 99.998%
Iron, foil, 8mm disks, thickness 0.125mm, hard, 99.5%
Iron, foil, 8mm disks, thickness 0.05mm, hard, 99.5%
Iron, foil, light tested, 100x100mm, thickness 0.025mm, hard, 99.5%
Iron, rod, 1000mm, diameter 2.0mm, as drawn, 99.99+%
Iron, foil, not light tested, 100x100mm, thickness 0.007mm, 99.85%
Iron, foil, light tested, 100x100mm, thickness 0.038mm, hard, 99.5%
Iron, foil, not light tested, 100x100mm, thickness 0.003mm, 99.85%
Iron, rod, 100mm, diameter 25mm, as drawn, 98+%