Merck
  • Comparison of two Gram stain point-of-care systems for urogenital gonorrhoea among high-risk patients: diagnostic accuracy and cost-effectiveness before and after changing the screening algorithm at an STI clinic in Amsterdam.
Comparison of two Gram stain point-of-care systems for urogenital gonorrhoea among high-risk patients: diagnostic accuracy and cost-effectiveness before and after changing the screening algorithm at an STI clinic in Amsterdam.
Sexually transmitted infections (2014-05-27)
M Bartelsman, M Straetemans, K Vaughan, S Alba, M S van Rooijen, W R Faber, H J C de Vries
ABSTRACT
To compare point-of-care (POC) systems in two different periods: (1) before 2010 when all high-risk patients were offered POC management for urogenital gonorrhoea by Gram stain examination; and (2) after 2010 when only those with symptoms were offered Gram stain examination. Retrospective comparison of a Gram stain POC system to all high-risk patients (2008-2009) with only those with urogenital symptoms (2010-2011) on diagnostic accuracy, loss to follow-up, presumptively and correctly treated infections and diagnostic costs. Culture was the reference diagnostic method. In men the sensitivity of the Gram stain was 95.9% (95% CI 93.1% to 97.8%) in 2008-2009 and 95.4% (95% CI 93.7% to 96.8%) in 2010-2011, and in women the sensitivity was 32.0% (95% CI 19.5% to 46.7%) and 23.1% (95% CI 16.1% to 31.3%), respectively. In both periods the overall specificity was high (99.9% (95% CI 99.8% to 100%) and 99.8% (95% CI 99.7% to 99.9%), respectively). The positive predictive value (PPV) and negative predictive value (NPV) before and after 2010 were also high: PPV 97.0% (95% CI 94.5% to 98.5%) and 97.7% (95% CI 96.3% to 98.6%), respectively; NPV 99.6% (95% CI 99.4% to 99.7%) and 98.8% (95% CI 98.5% to 99.0%), respectively. There were no differences between the two time periods in loss to follow-up (7.1% vs 7.0%). Offering Gram stains only to symptomatic high-risk patients as opposed to all high-risk patients saved €2.34 per correctly managed consultation (a reduction of 7.7%). The sensitivity of the Gram stain is high in men but low in women. When offered only to high-risk patients with urogenital symptoms, the cost per correctly managed consultation is reduced by 7.7% without a significant difference in accuracy and loss to follow-up.
Product Description
Supelco
Azithromycin, Pharmaceutical Secondary Standard; Certified Reference Material
Azithromycin for peak identification, European Pharmacopoeia (EP) Reference Standard
USP
Azithromycin Identity, United States Pharmacopeia (USP) Reference Standard
Azithromycin, European Pharmacopoeia (EP) Reference Standard
Azithromycin for system suitability, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Gentian Violet, meets USP testing specifications
USP
Azithromycin, United States Pharmacopeia (USP) Reference Standard
Supelco
Ceftriaxone Sodium, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Crystal Violet, VETRANAL®, analytical standard
Sigma-Aldrich
Crystal Violet, indicator for the determination of the redox potential, S. No.: 785
Sigma-Aldrich
Crystal Violet, for microscopy (Bact., Bot., Hist., Vit.), indicator (pH 0.1-2.0)
Sigma-Aldrich
Azithromycin
Sigma-Aldrich
Ceftriaxone disodium salt hemi(heptahydrate), third-generation cephalosporin antibiotic
Sigma-Aldrich
Crystal Violet, certified by the Biological Stain Commission
Sigma-Aldrich
Crystal Violet, ACS reagent, ≥90.0% anhydrous basis
Sigma-Aldrich
Azithromycin dihydrate, ≥98% (HPLC)
USP
Ceftriaxone sodium, United States Pharmacopeia (USP) Reference Standard
Ceftriaxone sodium, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Crystal Violet Solution, 1%, aqueous solution
Methylrosanilinium chloride, European Pharmacopoeia (EP) Reference Standard
Methylrosanilinium for system suitability, European Pharmacopoeia (EP) Reference Standard