Merck
  • Burkholderia vietnamiensis C09V as the functional biomaterial used to remove crystal violet and Cu(II).
Burkholderia vietnamiensis C09V as the functional biomaterial used to remove crystal violet and Cu(II).
Ecotoxicology and environmental safety (2014-05-02)
Fengfei Zhou, Ying Cheng, Li Gan, Zuliang Chen, Mallavarapu Megharaj, Ravendra Naidu
ABSTRACT
Burkholderia vietnamiensis C09V (B.V. C09V) was used to remove both crystal violet (CV) and Cu(II) because dye effluents often contain dyes and metal ions. Inhibiting the strain׳s growth through the biosorption of Cu(II) on B.V. C09V and promoting its growth by using CV as a carbon source led to the degradation of CV (30mg/L). It fell to 36.9 percent and the amount of Cu(II) (50mg/L) removed rose to 34.9 percent in the presence of both CV and Cu(II). This outcome is comparable to the single presence of CV and Cu(II). EDS analysis showed that Cu(II) was adsorbed onto the strain (the atomic percentage of Cu(II) was 1.9 percent), while kinetic studies indicated that firstly, the decolorization of CV fitted well to the pseudo first-order degradation kinetic model and secondly, the biosorption of Cu(II) fitted well to the pseudo second-order kinetic model. The degradation rate constants of CV were stable in the 0.101-0.0068/h range and R(2) was both higher than 0.981 when Cu(II) concentrations were present. Furthermore, the biosorption capacity of Cu(II) ranged from 38.8 to 20.3mg/g at the CV concentration of 30mg/L (both R(2)>0.96). This suggests that the strain has the potential to degrade CV and facilitate the biosorption of Cu(II) in dye effluent.
Sigma-Aldrich
Gentian Violet, meets USP testing specifications
Sigma-Aldrich
Crystal Violet Solution, 1%, aqueous solution
Sigma-Aldrich
Crystal Violet, S. No.: 785
Sigma-Aldrich
Crystal Violet, certified by the Biological Stain Commission
Sigma-Aldrich
Crystal Violet, for microscopy (Bact., Bot., Hist., Vit.), indicator (pH 0.1-2.0)
Sigma-Aldrich
Crystal Violet, ACS reagent, ≥90.0% anhydrous basis
Supelco
Crystal Violet, VETRANAL®, analytical standard
Methylrosanilinium for system suitability, European Pharmacopoeia (EP) Reference Standard
Methylrosanilinium chloride, European Pharmacopoeia (EP) Reference Standard