Merck
  • Optimization of self-nanoemulsifying systems for the enhancement of in vivo hypoglycemic efficacy of glimepiride transdermal patches.
Optimization of self-nanoemulsifying systems for the enhancement of in vivo hypoglycemic efficacy of glimepiride transdermal patches.
Expert opinion on drug delivery (2014-04-08)
Osama A A Ahmed, Mohsen I Afouna, Khalid M El-Say, Ashraf B Abdel-Naim, Alaa Khedr, Zainy M Banjar
ABSTRACT
To optimize and use of glimepiride (GMD)-loaded self-nanoemulsifying delivery systems (SNEDs) for the preparation of transdermal patches. Mixture design was utilized to optimize GMD-loaded SNEDs in acidic and aqueous pH media. Optimized GMD-loaded SNEDs were used in the preparation of chitosan (acidic) and hydroxypropyl methyl cellulose (HPMC) (aqueous) films. The prepared optimized formulations were investigated for ex vivo skin permeation, for in vivo hypoglycemic activity and for their pharmacokinetic parameters using animal model. The optimized formulations showed flux value of (2.88 and 4.428 μg/cm(2)/h) through rat skin for chitosan and HPMC films, respectively. The pattern of GMD release from both formulations was in favor of Higuchi and approaching zero order models. The n values for Korsmeyer-Peppas equation were characteristic of anomalous (non-Fickian) release mechanism. Moreover, HPMC patches have shown significant reductions (p < 0.05) in blood glucose levels; (213.33 ± 15.19) mg/100 ml from the base-line measurement after 12 h of application. Optimized GMD SNEDs patches were found to improve GMD skin permeability and the essential pharmacokinetic parameters. Further extensive pre/clinical studies are necessary prior to use transdermal GMD as a valuable alternative to peroral dosage forms with improved bioavailability, longer duration of action and more patient convenience.
Sigma-Aldrich
Methyl cellulose, viscosity 3000-5500 mPa.s, 2 % in H2O(20 °C)
Sigma-Aldrich
Methyl cellulose, 27.5-31.5% methoxyl basis
Sigma-Aldrich
Methyl cellulose, tested according to Ph. Eur.
Sigma-Aldrich
(Hydroxypropyl)methyl cellulose, average Mn ~86,000
Sigma-Aldrich
Methyl cellulose, medium viscosity, Methoxyl content 27.5-31.5 %
Sigma-Aldrich
Chitosan, high molecular weight
Sigma-Aldrich
Chitosan, low molecular weight
Sigma-Aldrich
Chitosan, from shrimp shells, practical grade
Sigma-Aldrich
Methyl cellulose, viscosity: 1,500 cP
Sigma-Aldrich
Methyl cellulose, viscosity: 4,000 cP
Sigma-Aldrich
Methyl cellulose, viscosity: 25 cP
Sigma-Aldrich
Methyl cellulose, viscosity: 15 cP
Sigma-Aldrich
Methyl cellulose, viscosity: 15 cP, BioReagent, suitable for cell culture
Sigma-Aldrich
Methyl cellulose, 27.5-31.5% (Methoxyl content), viscosity: 400 cP
Sigma-Aldrich
Methyl cellulose, 26.0-33.0% (Methoxy group (dry basis)), meets USP testing specifications, viscosity: 1,500 cP
Sigma-Aldrich
Chitosan, from shrimp shells, ≥75% (deacetylated)
Sigma-Aldrich
Methyl cellulose, 26.0-33.0% (methoxyl group, on Dry Basis), meets USP testing specifications, viscosity: 400 cP
USP
Glimepiride, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Glimepiride, ≥98% (HPLC), solid
Sigma-Aldrich
Chitosan, medium molecular weight
Glimepiride, European Pharmacopoeia (EP) Reference Standard
Glimepiride for system suitability, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Chitosan from shrimp shells, low-viscous