Merck
  • In vitro toxicity of nanoceria: effect of coating and stability in biofluids.
In vitro toxicity of nanoceria: effect of coating and stability in biofluids.
Nanotoxicology (2013-08-07)
Nawel Ould-Moussa, Malak Safi, Marie-Alice Guedeau-Boudeville, David Montero, Hélène Conjeaud, Jean-François Berret
ABSTRACT
Due to the increasing use of nanometric cerium oxide in applications, concerns about the toxicity of these particles have been raised and have resulted in a large number of studies. We report here on the interactions between 7 nm anionically charged cerium oxide particles and living mammalian cells. By a modification of the particle coating including low-molecular weight ligands and polymers, two generic behaviours are compared: particles coated with citrate ions that precipitate in biofluids and particles coated with poly(acrylic acid) that are stable and remain nanometric. We find that nanoceria covered with both coating agents are taken up by mouse fibroblasts and localized into membrane-bound compartments. However, flow cytometry and electron microscopy reveal that as a result of their precipitation, citrate-coated particles interact more strongly with cells. At cerium concentration above 1 mM, only citrate-coated nanoceria (and not particles coated with poly(acrylic acid)) display toxicity and moderate genotoxicity. The results demonstrate that the control of the surface chemistry of the particles and its ability to prevent aggregation can affect the toxicity of nanomaterials.
Sigma-Aldrich
Cerium(IV) oxide, nanopowder, <50 nm particle size (BET), 99.95% trace rare earth metals basis
Sigma-Aldrich
Cerium, chips
Sigma-Aldrich
Cerium(IV) oxide, dispersion, 20 wt. % colloidal dispersion in 2.5% acetic acid, 30-50 nm avg. part. size
Sigma-Aldrich
Cerium(IV) oxide, powder, <5 μm, 99.9% trace metals basis
Sigma-Aldrich
Cerium(IV) oxide, nanopowder, <25 nm particle size (BET)
Sigma-Aldrich
Cerium, ingot, under oil, 99.9% trace rare earth metals basis
Sigma-Aldrich
Cerium(IV) oxide, powder, 99.995% trace metals basis
Sigma-Aldrich
Cerium(IV) oxide, ≥99.0%
Sigma-Aldrich
Cerium(IV) oxide, dispersion, nanoparticles, <25 nm particle size, 10 wt. % in H2O
Cerium, foil, not light tested, 25x25mm, thickness 0.025mm, as rolled, 99.9%
Cerium, foil, 25x25mm, thickness 0.1mm, as rolled, 99.9%
Cerium, foil, not light tested, 50x50mm, thickness 0.025mm, as rolled, 99.9%
Cerium, rod, 50mm, diameter 6.35mm, cast, 99.9%
Cerium, foil, 50x50mm, thickness 0.1mm, as rolled, 99.9%
Cerium, wire reel, 50mm, diameter 1.0mm, hard, 99.9%
Cerium, wire reel, 100mm, diameter 1.0mm, hard, 99.9%
Cerium, rod, 100mm, diameter 12.7mm, cast, 99.9%
Cerium, wire reel, 25mm, diameter 1.0mm, hard, 99.9%
Cerium, rod, 50mm, diameter 12.7mm, cast, 99.9%
Cerium, rod, 100mm, diameter 6.35mm, cast, 99.9%
Cerium, foil, not light tested, 25x25mm, thickness 0.005mm, as rolled, 99.9%
Cerium, foil, light tested, 25x25mm, thickness 0.25mm, as rolled, 99.9%