Merck
  • Uptake and toxicity of copper oxide nanoparticles in cultured primary brain astrocytes.
Uptake and toxicity of copper oxide nanoparticles in cultured primary brain astrocytes.
Nanotoxicology (2013-07-31)
Felix Bulcke, Karsten Thiel, Ralf Dringen
ABSTRACT
To test for consequences of an exposure of brain cells to copper oxide nanoparticles (CuO-NPs), we synthesised and characterised dimercaptosuccinate-coated CuO-NPs. These particles had a diameter of around 5 nm as determined by transmission electron microscopy, while their average hydrodynamic diameter in aqueous dispersion was 136 ± 4 nm. Dispersion in cell-culture medium containing 10% fetal calf serum increased the hydrodynamic diameter to 178 ± 12 nm and shifted the zeta potential of the particles from -49 ± 7 mV (in water) to -10 ± 3 mV. Exposure of cultured primary brain astrocytes to CuO-NPs increased the cellular copper levels and compromised the cell viability in a time-, concentration- and temperature-dependent manner. Application of CuO-NPs in concentrations above 100 µM copper (6.4 µg/ml) severely compromised the viability of the cells, as demonstrated by a lowered 3-(4,5-dimethylthiazol-2-yl)-2,5-diphenyltetrazolium bromide reduction capacity, a lowered cellular lactate dehydrogenase activity and an increased membrane permeability for the fluorescent dye propidium iodide. Copper internalisation as well as cell toxicity of astrocytes exposed to CuO-NPs were similar to that observed for cells that had been incubated with copper salts. The CuO-NP-induced toxicity was accompanied by an increase in the generation of reactive oxygen species (ROS) in the cells. Both, ROS formation and cell toxicity in CuO-NP-treated astrocytes, were lowered in the presence of the cell-permeable copper chelator tetrathiomolybdate. These data demonstrate that CuO-NPs are taken up by cultured astrocytes and suggest that excess of internalised CuO-NPs cause cell toxicity by accelerating the formation of ROS.
Sigma-Aldrich
Molybdenum, powder, 1-5 μm, ≥99.9% trace metals basis
Sigma-Aldrich
Molybdenum, powder, <150 μm, 99.99% trace metals basis
Sigma-Aldrich
Molybdenum, foil, thickness 1.0 mm, ≥99.9% trace metals basis
Sigma-Aldrich
Copper(II) oxide, 99.999% trace metals basis
Sigma-Aldrich
Molybdenum, powder, <150 μm, 99.9% trace metals basis
Sigma-Aldrich
Molybdenum, foil, thickness 0.1 mm, ≥99.9% trace metals basis
Sigma-Aldrich
Molybdenum, foil, thickness 0.025 mm, ≥99.9% trace metals basis
Sigma-Aldrich
Copper(II) oxide, nanopowder, <50 nm particle size (TEM)
Sigma-Aldrich
Molybdenum, nanopowder, <100 nm particle size (TEM), 99.8% trace metals basis
Sigma-Aldrich
Copper(II) oxide, powder, 99.99% trace metals basis
Sigma-Aldrich
Copper(II) oxide, powder, 99.995% trace metals basis
Sigma-Aldrich
Molybdenum, powder, 10 μm, ≥99.95% trace metals basis
Molybdenum, foil, 25mm disks, thickness 0.015mm, annealed, 99.9%
Molybdenum, foil, 10mm disks, thickness 0.05mm, annealed, 99.9%
Molybdenum, foil, 10m coil, thickness 0.04mm, coil width 0.7mm, annealed, 99.9%
Molybdenum, foil, 25mm disks, thickness 0.15mm, annealed, 99.9%
Molybdenum, foil, 1m coil, thickness 0.01mm, 99.9%
Molybdenum, foil, 10mm disks, thickness 0.008mm, 99.9%
Molybdenum, foil, not light tested, 25x25mm, thickness 0.005mm, 99.9%
Molybdenum, foil, light tested, 25x25mm, thickness 0.05mm, annealed, 99.9%
Sigma-Aldrich
Copper(II) oxide on alumina, 14-30 mesh, extent of labeling: 13 wt. % loading
Molybdenum, rod, 500mm, diameter 1.5mm, centerless ground, 99.9%
Molybdenum, tube, 500mm, outside diameter 1.5mm, inside diameter 0.95mm, wall thickness 0.275mm, 99.9%
Molybdenum, tube, 50mm, outside diameter 6.4mm, inside diameter 4.6mm, wall thickness 0.9mm, 99.9%
Molybdenum, rod, 500mm, diameter 12mm, centerless ground, 99.9%
Molybdenum, rod, 100mm, diameter 25mm, centerless ground, 99.9%
Molybdenum, rod, 100mm, diameter 4.0mm, centerless ground, 99.9%
Molybdenum, rod, 500mm, diameter 2.0mm, centerless ground, 99.9%
Molybdenum, rod, 500mm, diameter 15.0mm, centerless ground, 99.9%
Molybdenum, rod, 50mm, diameter 5.0mm, 99.98%