Merck
  • Mometasone furoate hydrogel for scalp use: in vitro and in vivo evaluation.
Mometasone furoate hydrogel for scalp use: in vitro and in vivo evaluation.
Pharmaceutical development and technology (2013-07-23)
Ana Salgado, Sara Raposo, Joana Marto, Alexandra N Silva, Sandra Simões, Helena M Ribeiro
ABSTRACT
Dermatological inflammatory diseases often affect the scalp and the eyebrows. Common dosage forms available on the market for those situations are lotions; however, the presence of hair limits their use. Gels, for their consistency and adhesiveness, are a suitable alternative to the lotions in these situations. The aim of this study was to develop a new stable gel containing mometasone furoate (MF), with anti-inflammatory activity and a controlled delivery, to improve topical treatment of scalp dermatitis. Pharmaceutical development, physical and chemical characterization, stability, in vitro release and permeation studies and in vivo anti-inflammatory activity were performed. The gel presented an acidic pH and an apparent viscosity of 35 Pa.s. The microbiological analysis showed that the results were within the established specification limits. The release and the permeation profiles suggest that the drug is mainly retained in the upper skin layers. MF gel was tested in an animal model of cutaneous inflammation and presented similar anti-inflammatory activity compared to a commercially available MF dosage form. The gel was chemically, physically and microbiologically stable. The results suggest that the developed hydrogel formulation containing MF can be of actual value for improving the clinical effectiveness in the treatment of scalp dermatitis.
Sigma-Aldrich
Methyl cellulose, 27.5-31.5% (Methoxyl content), viscosity: 400 cP
Sigma-Aldrich
Methyl cellulose, 26.0-33.0% (methoxyl group, on Dry Basis), meets USP testing specifications, viscosity: 400 cP
Sigma-Aldrich
Methyl cellulose, viscosity: 15 cP, BioReagent, suitable for cell culture
Sigma-Aldrich
Methyl cellulose, viscosity: 25 cP
Sigma-Aldrich
Methyl cellulose, viscosity: 15 cP
Sigma-Aldrich
Methyl cellulose, viscosity: 4,000 cP
Sigma-Aldrich
Methyl cellulose, viscosity: 1,500 cP
Sigma-Aldrich
Methyl cellulose, 26.0-33.0% (Methoxy group (dry basis)), meets USP testing specifications, viscosity: 1,500 cP
Sigma-Aldrich
Methyl cellulose, tested according to Ph. Eur.
Sigma-Aldrich
Methyl cellulose, 27.5-31.5% methoxyl basis
Sigma-Aldrich
Methyl cellulose, medium viscosity, Methoxyl content 27.5-31.5 %
Sigma-Aldrich
Poly(ethylene glycol) dimethacrylate, average Mn 750, contains 900-1100 ppm MEHQ as inhibitor
Sigma-Aldrich
Poly(ethylene glycol) dimethacrylate, average Mn 6,000, contains 1000 ppm 4-methoxyphenol as inhibitor
Sigma-Aldrich
Methyl cellulose, viscosity 3000-5500 mPa.s, 2 % in H2O(20 °C)
Sigma-Aldrich
Poly(ethylene glycol) dimethacrylate, average Mn 20,000, contains MEHQ as inhibitor
Sigma-Aldrich
Poly(ethylene glycol) dimethacrylate, average Mn 10,000, contains MEHQ as inhibitor
Sigma-Aldrich
Poly(ethylene glycol) dimethacrylate, average Mn 2000, contains ~1000 ppm MeHQ as stabilizer
Sigma-Aldrich
Poly(ethylene glycol) dimethacrylate, average Mn 550, contains 80-120 ppm MEHQ as inhibitor, 270-330 ppm BHT as inhibitor
Sigma-Aldrich
Mometasone furoate, ≥98% (HPLC)
Mometasone furoate, European Pharmacopoeia (EP) Reference Standard
USP
Mometasone furoate, United States Pharmacopeia (USP) Reference Standard
Supelco
Mometasone Furoate, Pharmaceutical Secondary Standard; Certified Reference Material