Merck
A role for oleoylethanolamide in chronic lymphocytic leukemia.
Leukemia (2014-01-15)
M Masoodi, E Lee, M Eiden, A Bahlo, Y Shi, R B Ceddia, C Baccei, P Prasit, D E Spaner
ABSTRACT
Oleoylethanolamide (OEA) is a bioactive lipid that stimulates nuclear and G protein-coupled receptors and regulates appetite and fat metabolism. It has not previously been shown to have a role in cancer. However, a mass spectrometry-based lipidomics platform revealed the presence of high amounts of OEA in the plasma of chronic lymphocytic leukemia (CLL) patients compared with normal donors. CLL cells produced OEA and the magnitude of plasma OEA levels was related directly to the circulating leukemic cell number. OEA from CLL cells was increased by URB-597, an inhibitor of fatty acid amide hydrolase (FAAH), and decreased by inflammatory mediators that downregulate expression of N-acylphosphatidylethanolamine-specific phospholipase D (NAPE-PLD). These enzymes degrade and synthesize OEA, respectively. Nonphysiologic doses of OEA prevented spontaneous apoptosis of CLL cells in a receptor-independent manner that was mimicked by its free fatty acid (FFA) derivative oleate. However, OEA-containing supernatants from CLL cells induced lipolysis in adipocytes, lipid products from adipocytes protected CLL cells from cytotoxic chemotherapy, and increased levels of FFAs were found in CLL plasma that correlated with OEA. We suggest OEA is a lipolytic factor produced by CLL cells to fuel their growth with a potential role in drug resistance and cancer cachexia.
Product Description
Supelco
Dehydrated Alcohol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, NMR reference standard
Supelco
Cholesterol solution, certified reference material, 10 mg/mL in chloroform
Sigma-Aldrich
SyntheChol® NS0 Supplement, 500 ×, synthetic cholesterol, animal component-free, aqueous solution, sterile-filtered, suitable for cell culture
Supelco
Ethanol solution, certified reference material, 2000 μg/mL in methanol
Sigma-Aldrich
Resiquimod, ≥98% (HPLC)
USP
Fenofibrate, United States Pharmacopeia (USP) Reference Standard
Fenofibrate, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Cholesterol, powder, BioReagent, suitable for cell culture, ≥99%
Sigma-Aldrich
Fenofibrate, ≥99%, powder
Sigma-Aldrich
Cholesterol, from sheep wool, ≥92.5% (GC), powder
Sigma-Aldrich
Cholesterol, Sigma Grade, ≥99%
Sigma-Aldrich
Chlorambucil
Sigma-Aldrich
7-Aminoactinomycin D, ~97% (HPLC), powder
Supelco
Acetic acid, analytical standard
Supelco
Methanol, analytical standard
Supelco
Methyl formate, analytical standard
Sigma-Aldrich
Cholesterol, tested according to Ph. Eur.
Sigma-Aldrich
Acetic acid, for luminescence, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Cholesterol, from lanolin, ≥99.0% (GC)
Sigma-Aldrich
Methyl formate, anhydrous, 99%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Chlorambucil, European Pharmacopoeia (EP) Reference Standard
USP
Dehydrated Alcohol, United States Pharmacopeia (USP) Reference Standard
Supelco
Fenofibrate, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
Acetic acid-12C2, 99.9 atom % 12C
Sigma-Aldrich
Methanol, puriss., meets analytical specification of Ph Eur, ≥99.7% (GC)