Merck
  • Promotion of proton conduction in polymer electrolyte membranes by 1H-1,2,3-triazole.
Promotion of proton conduction in polymer electrolyte membranes by 1H-1,2,3-triazole.
Journal of the American Chemical Society (2005-08-04)
Zhen Zhou, Siwen Li, Yuelan Zhang, Meilin Liu, Wen Li
ABSTRACT
We report 1H-1,2,3-triazole as an active group to dramatically enhance proton conduction in a polymer electrolyte membrane (PEM). The conductivities of a poly(4-vinyl-1H-1,2,3-triazole) membrane without any acidic dopants are about 105 times greater than those of poly(4-vinylimidazole) in dry air at 50-150 degrees C. Polymers with groups promoting proton conduction attached to the backbone have great potential to offer excellent mechanical properties and long-term stability. Further, 1H-1,2,3-triazole and PEMs containing 1H-1,2,3-triazole are stable in a wide potential range, implying excellent electrochemical stability under fuel cell operating conditions.
Sigma-Aldrich
1H-1,2,3-Triazole, 97%