Merck
  • Effect of a glutathione S-transferase inhibitor on oxidative stress and ischemia-reperfusion-induced apoptotic signalling of cultured cardiomyocytes.
Effect of a glutathione S-transferase inhibitor on oxidative stress and ischemia-reperfusion-induced apoptotic signalling of cultured cardiomyocytes.
Experimental and clinical cardiology (2011-11-09)
E Röth, N Marczin, B Balatonyi, S Ghosh, V Kovács, N Alotti, B Borsiczky, B Gasz
ABSTRACT
Oxidative stress and ischemia-reperfusion (I/R) injury are crucial in the pathogenesis of cardiovascular diseases. The antioxidant glutathione S-transferase (GST) is responsible for the high-capacity metabolic inactivation of electrophilic compounds and toxic substrates. The main objective of the present study was to examine the effect of GST inhibition (with the administration of ethacrynic acid [EA]) on the viability and apoptosis of cardiomyocytes when these cells are exposed to various stress components of I/R and mitogen-activated protein kinase (c-Jun N-terminal kinase, p38 and extracellular signal-regulated kinase [ERK]) inhibitors. The primary culture of neonatal rat cardiomyocytes was divided into six experimental groups: control group of cells (group 1), cells exposed to H(2)O(2) (group 2), I/R (group 3), I/R and EA (group 4), H(2)O(2) coupled with EA (group 5), and EA alone (group 6). The viability of cardiomyocytes was determined using a colorimetric MTT assay. The apoptosis ratio was evaluated via fluorescein isothiocyanate-labelled annexin V and propidium iodide staining. c-Jun N-terminal kinase, p38, Akt/protein kinase B and ERK/p42-p44 transcription factors were monitored with flow cytometry. c-Jun N-terminal kinase activation increased due to GST inhibition during I/R. EA administration led to a significant increase in p38 activation following both H(2)O(2) treatment and I/R. ERK phosphorylation increased when GST was exposed to I/R. A pronounced decrease in Akt phosphorylation was observed when cells were cotreated with EA and H(2)O(2). GST plays an important role as a regulator of mitogen-activated protein kinase pathways in I/R injury.
Sigma-Aldrich
Ethacrynic acid, ≥97% (HPLC)
Pricing and availability is not currently available.