Merck
  • Effect of changing the fine particle mass of inhaled beclomethasone dipropionate on intrapulmonary deposition and pharmacokinetics.
Effect of changing the fine particle mass of inhaled beclomethasone dipropionate on intrapulmonary deposition and pharmacokinetics.
Respiratory medicine (1998-12-16)
J P Seale, L I Harrison
ABSTRACT
Reformulation of beclomethasone dipropionate (BDP) in the chlorofluorocarbon (CFC)-free propellant hydrofluoroalkane-134a (HFA) gave the opportunity to produce a solution formulation that provides a greater total mass of fine drug particles than the current CFC suspension metered dose inhaler (MDI). The HFA-BDP MDI was studied in three pharmacokinetic trials in asthmatic patients. Serum levels of BDP plus metabolites [total beclomethasone (total BOH) assay] were used to test whether the increased fine particle mass of HFA-BDP would result in improved intrapulmonary deposition and subsequent differences in serum profiles. Serum levels, maximum serum concentrations and area under the serum concentration-time curves of total BOH following both single and multiple doses of HFA-BDP were similar to those obtained with approximately twice the dose of CFC-BDP. The observed lower bioavailability of CFC-BDP compared with HFA-BDP could be explained if most of each inhaled dose from the CFC-BDP MDI was swallowed and absorbed from the gastrointestinal tract, while most of each inhaled dose from the HFA-BDP MDI was absorbed from the lungs. Deposition studies have confirmed this explanation. These results suggest that asthmatic patients can be treated with lower total daily doses of drug from HFA-BDP extrafine aerosol than from CFC-BDP.
Product Description
Supelco
Beclomethasone dipropionate, analytical standard, for drug analysis
Beclomethasone dipropionate, anhydrous, European Pharmacopoeia (EP) Reference Standard
USP
Beclomethasone dipropionate, United States Pharmacopeia (USP) Reference Standard
Beclometasone dipropionate for peak identification, European Pharmacopoeia (EP) Reference Standard
Beclometasone dipropionate for system suitability, European Pharmacopoeia (EP) Reference Standard