Merck
Bilateral renal cortical necrosis associated with cefuroxime axetil.
Clinical nephrology (1998-05-16)
H J Manley, G R Bailie, G Eisele
ABSTRACT
Cefuroxime axetil has been associated with few reported adverse effects. We report a case of bilateral renal cortical necrosis in a female after receiving 7 doses over 4 treatment days. The patient presented with worsening symptoms consisting of arthralgias, pruritus, and abdominal pain. Laboratory data obtained was indicative of worsening renal failure and thrombocytopenia. The patient required hemodialysis by the third day. Kidney biopsy revealed cortical necrosis. The possible pathogenesis of cefuroxime axetil causing cortical necrosis in this case and a review of other reported cases of chemical induced renal cortical necrosis is discussed.
Product Description
USP
Cefuroxime Axetil, United States Pharmacopeia (USP) Reference Standard
Cefuroxime Axetil, European Pharmacopoeia (EP) Reference Standard
Supelco
Cefuroxime, VETRANAL®, analytical standard