Merck
Nystatin prophylaxis in immunocompromised children.
The Annals of pharmacotherapy (1996-05-01)
T L Taylor
ABSTRACT
In conclusion, the use of nystatin for fungal prophylaxis in hematology/oncology/bone marrow transplantation patients appears to convey little or no benefit over no therapy. Nystatin treatment significantly reduced the frequency of multiple-site colonization and of persistently positive oropharyngeal cultures, but was not able to prevent the occurrence of disseminated fungal infections. Other antifungal agents (e.g., ketoconazole, fluconazole, itraconazole, oral or intravenous amphotericin B) may be of benefit. Studies comparing nystatin with other agents are available in the literature but are beyond the scope of this review.
Sigma-Aldrich
Nystatin, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Nystatin, powder, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
Nystatin, ≥4,400 units/mg
USP
Nystatin, United States Pharmacopeia (USP) Reference Standard
Nystatin, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Nystatin Suspension, suspension, 10,000 unit/mL in DPBS, BioReagent, suitable for cell culture