Merck
  • Biochemical and molecular aspects of mammalian susceptibility to aflatoxin B1 carcinogenicity.
Biochemical and molecular aspects of mammalian susceptibility to aflatoxin B1 carcinogenicity.
Proceedings of the Society for Experimental Biology and Medicine. Society for Experimental Biology and Medicine (New York, N.Y.) (1995-03-01)
T E Massey, R K Stewart, J M Daniels, L Liu
ABSTRACT
Aflatoxin B1 (AFB1) is a fungal toxin that has been implicated as a causative agent in human hepatic and extrahepatic carcinogenesis. In this review, the mechanisms involved in AFB1 toxicity are delineated, in order to describe the features that make a specific cell, tissue, or species susceptible to the mycotoxin. Important considerations include: (i) different mechanisms for bioactivation of AFB1 to its ultimate carcinogenic epoxide metabolite; (ii) the balance between bioactivation to and detoxification of the epoxide; (iii) the interaction of AFB1 epoxide with DNA and the mutational events leading to neoplastic transformation; (iv) the role of cytotoxicity in AFB1 carcinogenesis; (v) the significance of nonepoxide metabolites in toxicity; and (vi) the contribution of mycotoxin-unrelated disease processes. Although considerable controversy remains about the importance of specific events, a great deal has been learned about biochemical and molecular actions of AFB1.
Sigma-Aldrich
Aflatoxin B1 from Aspergillus flavus, from Aspergillus flavus
Supelco
Aflatoxin B1 solution, 3 μg/mL in benzene:acetonitrile (98:2), analytical standard
Aflatoxin B1 solution, 3.79 μg/g in acetonitrile, ERM®, certified reference material
Supelco
Aflatoxin B1 solution, 2 μg/mL in acetonitrile, analytical standard