Merck
  • L-thyroxine overdose: a case of marked, severe, prolonged, excess ingestion and review of the literature.
L-thyroxine overdose: a case of marked, severe, prolonged, excess ingestion and review of the literature.
Thyroidology (1993-08-01)
B Shapiro, M D Gross, O Geatti
ABSTRACT
A case of a man with thyroid cancer who ingested between 0.9 and 3.3 mg l-thyroxine per day for over 10 years (the highest dose for 3 years) is reported. This had been prescribed for suppression of TSH for a well differentiated thyroid cancer. He was essentially asymptomatic and suffered no apparent ill effects from this prolonged and markedly excessive dosage of l-thyroxine. The literature lists a wide range of ill effects from both chronic and acute thyroid hormone overdosage but also records many examples of tolerance to excessive levels of exogenous thyroid hormone. The various circumstances leading to thyroid hormone overdose and potential ill-effects are reviewed.
Sigma-Aldrich
L-Thyroxine, ≥98% (HPLC)
Sigma-Aldrich
L-Thyroxine, powder, BioReagent, suitable for cell culture
USP
Levothyroxine, United States Pharmacopeia (USP) Reference Standard