Merck
Clinical safety of aspartame.
American family physician (1989-02-01)
D A Yost
ABSTRACT
Aspartame is a synthetic sweetener commonly used in soft drinks and many foods. Even with high doses, the metabolites of this sweetener do not accumulate in toxic amounts. To date, no definite symptom complex has been connected with aspartame, and it is considered safe for use in all populations, including diabetics, phenylketonuric heterozygotes and pregnant women.
Product Description
Supelco
Aspartame, Pharmaceutical Secondary Standard; Certified Reference Material
SKU
Pack Size
Availability
Price
Quantity
Supelco
Aspartame, analytical standard
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Asp-Phe methyl ester, ≥98%
SKU
Pack Size
Availability
Price
Quantity
Aspartame, European Pharmacopoeia (EP) Reference Standard
SKU
Pack Size
Availability
Price
Quantity
USP
Aspartame, United States Pharmacopeia (USP) Reference Standard