Merck
  • Quinolones and non-steroidal anti-inflammatory drugs interacting with copper(II), nickel(II), cobalt(II) and zinc(II): structural features, biological evaluation and perspectives.
Quinolones and non-steroidal anti-inflammatory drugs interacting with copper(II), nickel(II), cobalt(II) and zinc(II): structural features, biological evaluation and perspectives.
Dalton transactions (Cambridge, England : 2003) (2013-03-27)
George Psomas, Dimitris P Kessissoglou
ABSTRACT
The structural features of copper(II), nickel(II), cobalt(II) and zinc(II) complexes with the antimicrobial drugs quinolones and non-steroidal anti-inflammatory drugs (NSAIDs) as ligands are discussed. The binding properties of these complexes to biomolecules (calf-thymus DNA, bovine or human serum albumin) are presented and evaluated. The biological activity (antimicrobial, antioxidant and antiproliferative) of selected complexes is investigated. Further perspectives concerning the synthesis and the biological activity of novel complexes with quinolones or NSAIDs attractive to synthetic chemists, biochemists and/or biologists are presented.
Sigma-Aldrich
Cobalt, pieces, 99.5% trace metals basis
Sigma-Aldrich
Cobalt, Carbon coated magnetic, nanopowder, <50 nm particle size (TEM), ≥99%
Sigma-Aldrich
Cobalt, granular, 99.99% trace metals basis
Sigma-Aldrich
Cobalt, rod, diam. 5.0 mm, 99.95% trace metals basis
Sigma-Aldrich
Cobalt, powder, <150 μm, ≥99.9% trace metals basis
Sigma-Aldrich
Nickel, rod, diam. 6.35 mm, ≥99.99% trace metals basis
Sigma-Aldrich
Nickel, foil, thickness 0.5 mm, 99.98% trace metals basis
Sigma-Aldrich
Nickel, powder, <50 μm, 99.7% trace metals basis
Sigma-Aldrich
Nickel, wire, diam. 0.5 mm, ≥99.9% trace metals basis
Sigma-Aldrich
Cobalt, foil, thickness 0.1 mm, ≥99.99%
Sigma-Aldrich
Cobalt, foil, thickness 0.1 mm, 99.95% trace metals basis
Sigma-Aldrich
Cobalt, foil, thickness 1.0 mm, 99.95% trace metals basis
Sigma-Aldrich
Cobalt, powder, 2 μm particle size, 99.8% trace metals basis
Sigma-Aldrich
Nickel, nanopowder, <100 nm avg. part. size, ≥99% trace metals basis
Sigma-Aldrich
Nickel, foil, thickness 0.1 mm, 99.98% trace metals basis
Sigma-Aldrich
Cobalt, wire, diam. 1.0 mm, 99.995% trace metals basis
Nickel, foil, light tested, 100x100mm, thickness 0.075mm, annealed, 99%
Nickel, foil, not light tested, 100x100mm, thickness 0.0125mm, annealed, 99%
Nickel, foil, light tested, 50x50mm, thickness 0.03mm, 99.9%
Nickel, foil, light tested, 150x150mm, thickness 0.0125mm, annealed, 99%
Nickel, foil, light tested, 100x100mm, thickness 0.01mm, 99.95%
Nickel, foil, light tested, 50x50mm, thickness 0.05mm, annealed, 99%
Nickel, foil, light tested, 25x25mm, thickness 0.025mm, annealed, 99%
Nickel, foil, 25mm disks, thickness 0.25mm, annealed, 99%
Nickel, foil, not light tested, 150x150mm, thickness 0.0125mm, 99.9%
Cobalt, foil, 25mm disks, thickness 0.01mm, 99.9%
Nickel, foil, light tested, 50x50mm, thickness 0.01mm, 99.95%
Nickel, foil, light tested, 100x100mm, thickness 0.05mm, as rolled, 99.99+%
Nickel, foil, not light tested, 100x100mm, thickness 0.008mm, 99.95%
Nickel, foil, light tested, 150x150mm, thickness 0.0125mm, 99.9%