Merck
  • Practical recommendations for coaches and athletes: a meta-analysis of sodium bicarbonate use for athletic performance.
Practical recommendations for coaches and athletes: a meta-analysis of sodium bicarbonate use for athletic performance.
Journal of strength and conditioning research (2012-04-17)
Daniel J Peart, Jason C Siegler, Rebecca V Vince
ABSTRACT
Sodium bicarbonate (NaHCO3) is a buffering agent that is suggested to improve performance by promoting the efflux of hydrogen ions from working cells and tissues. Research surrounding its efficacy as an ergogenic aid is conflicting, making it difficult to draw conclusions as to its effectiveness for training and competition. This study performed a meta-analysis of relevant research articles to allow the development of concise practical recommendations for coaches and athletes. The overall effect size for the influence of NaHCO3 on performance was moderate, and was significantly lower for specifically trained as opposed to recreationally trained participants.
Product Description
Supelco
Sodium bicarbonate concentrate, 0.1 M NaHCO3 in water, eluent concentrate for IC
USP
Sodium bicarbonate, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Sodium bicarbonate, tested according to Ph. Eur.
Sigma-Aldrich
Sodium bicarbonate, powder, BioReagent, for molecular biology, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Sodium bicarbonate solution, solution (7.5%), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Sodium bicarbonate, BioXtra, 99.5-100.5%
Sigma-Aldrich
Sodium bicarbonate, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Sodium bicarbonate, ReagentPlus®, ≥99.5%, powder
Sigma-Aldrich
Sodium bicarbonate, ACS reagent, ≥99.7%
Sigma-Aldrich
Sodium bicarbonate, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, E500, 99.0-100.5%, powder
Sigma-Aldrich
Sodium hydrogencarbonate, −40-+140 mesh, ≥95%
Sigma-Aldrich
Sodium bicarbonate-12C, 99.9 atom % 12C