Merck
Ergocalciferol and cholecalciferol in CKD.
American journal of kidney diseases : the official journal of the National Kidney Foundation (2012-05-09)
Sagar U Nigwekar, Ishir Bhan, Ravi Thadhani
ABSTRACT
The development of chronic kidney disease (CKD) is accompanied by a progressive decrease in the ability to produce 1,25-dihydroxyvitamin D. Pharmacological replacement with active vitamin D therefore has been a cornerstone of secondary hyperparathyroidism therapy in the end-stage renal disease population treated by long-term dialysis. Recent evidence suggests that extrarenal conversion of 25-hydroxyvitamin D to 1,25-dihydroxyvitamin D may have significant biological roles beyond those traditionally ascribed to vitamin D. Furthermore, low 25-hydroxyvitamin D levels are common in patients with all stages of CKD. This article focuses on the role of nutritional vitamin D replacement in CKD and aims to review vitamin D biology and summarize the existing literature regarding nutritional vitamin D replacement in these populations. Based on the current state of the evidence, we provide suggestions for clinical practice and address areas of uncertainty that need further research.
Product Description
Supelco
Cholecalciferol (Vitamin D3), Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Cholecalciferol (D3), analytical standard
Sigma-Aldrich
Cholecalciferol, ≥98% (HPLC)
Sigma-Aldrich
Cholecalciferol, analytical standard
Sigma-Aldrich
Cholecalciferol, meets USP testing specifications
Cholecalciferol, European Pharmacopoeia (EP) Reference Standard
USP
Cholecalciferol, United States Pharmacopeia (USP) Reference Standard