Sigma-Aldrich
Bismuth, pieces, 1-12 mm, 99.999% trace metals basis
Pricing and availability is not currently available.
Sigma-Aldrich
Bismuth, granular, ≥99.99% trace metals basis
Pricing and availability is not currently available.
Sigma-Aldrich
Bismuth, beads, 1-5 mm, 99.999% trace metals basis
Pricing and availability is not currently available.
Sigma-Aldrich
Bismuth, powder, −100 mesh, 99% trace metals basis
Pricing and availability is not currently available.
Sigma-Aldrich
Bismuth, shot, 4-30 mesh, 99.9% trace metals basis
Pricing and availability is not currently available.
Sigma-Aldrich
Bismuth, powder, −100 mesh, ≥99.99% trace metals basis
Pricing and availability is not currently available.
Bismuth, foil, not light tested, 25x25mm, thickness 0.02mm, permanent polyester support, 99.97%
Pricing and availability is not currently available.
Bismuth, foil, not light tested, 50x50mm, thickness 0.002mm, permanent polyester support, 99.97%
Pricing and availability is not currently available.
Bismuth, foil, not light tested, 100x100mm, thickness 0.02mm, permanent polyester support, 99.97%
Pricing and availability is not currently available.
Bismuth, foil, 25x25mm, thickness 1.0mm, 99.97%
Pricing and availability is not currently available.
Bismuth, foil, not light tested, 100x100mm, thickness 0.01mm, permanent polyester support, 99.97%
Pricing and availability is not currently available.
Bismuth, foil, 25x25mm, thickness 0.5mm, 100%
Pricing and availability is not currently available.
Bismuth, foil, not light tested, 25x25mm, thickness 0.0025mm, permanent polyester support, 99.97%
Pricing and availability is not currently available.
Bismuth, foil, not light tested, 25x25mm, thickness 0.025mm, permanent polyester support, 99.97%
Pricing and availability is not currently available.
Bismuth, foil, not light tested, 50x50mm, thickness 0.005mm, permanent polyester support, 99.97%
Pricing and availability is not currently available.
Bismuth, foil, not light tested, 50x50mm, thickness 0.05mm, permanent polyester support, 99.97%
Pricing and availability is not currently available.
Bismuth, foil, not light tested, 100x100mm, thickness 0.05mm, permanent polyester support, 99.97%
Pricing and availability is not currently available.
Bismuth, foil, 25x25mm, thickness 2.0mm, 100%
Pricing and availability is not currently available.
Bismuth, foil, not light tested, 25x25mm, thickness 0.05mm, permanent polyester support, 99.97%
Pricing and availability is not currently available.
Bismuth, foil, light tested, 50x50mm, thickness 0.015mm, permanent polyester support, 99.97%
Pricing and availability is not currently available.
Bismuth, foil, not light tested, 50x50mm, thickness 0.001mm, permanent polyester support, 99.97%
Pricing and availability is not currently available.
Bismuth, foil, light tested, 50x50mm, thickness 0.01mm, permanent polyester support, 99.97%
Pricing and availability is not currently available.
Bismuth, foil, light tested, 25x25mm, thickness 0.05mm, permanent polyester support, 99.97%
Pricing and availability is not currently available.
Bismuth, foil, not light tested, 50x50mm, thickness 0.0025mm, permanent polyester support, 99.97%
Pricing and availability is not currently available.
Bismuth, foil, not light tested, 150x150mm, thickness 0.025mm, permanent polyester support, 99.97%
Pricing and availability is not currently available.
Bismuth, foil, not light tested, 150x150mm, thickness 0.05mm, permanent polyester support, 99.97%
Pricing and availability is not currently available.
Bismuth, foil, 50x50mm, thickness 0.25mm, 100%
Pricing and availability is not currently available.
Bismuth, foil, 50x50mm, thickness 1.0mm, 100%
Pricing and availability is not currently available.
Bismuth, foil, light tested, 50x50mm, thickness 0.02mm, permanent polyester support, 99.97%
Pricing and availability is not currently available.
Bismuth, foil, not light tested, 150x150mm, thickness 0.02mm, permanent polyester support, 99.97%
Pricing and availability is not currently available.