Merck
[The metal tantalum in orthopedic applications].
Sheng wu yi xue gong cheng xue za zhi = Journal of biomedical engineering = Shengwu yixue gongchengxue zazhi (2011-05-25)
Juncai Deng, Yue Wang
ABSTRACT
This paper describes the biological characteristics of the metal tantalum (Ts) and the application status of tantalum in artificial joints, bone necrosis, spine, defects of bone and other aspects of bone. The early clinical application results of tantalum in bone diseases were satisfactory, but it is necessary to do further study of tantalum in a deep going way, and further detailed comparison between the tantalum with other metals as orthopedics implants. The advantage of tantalum materials as orthopedic implants still needs to be verified by a great deal of clinical cases for a long period of time.
Product Description
Tantalum, foil, light tested, 25x25mm, thickness 0.005mm, 99.9%
Tantalum, foil, light tested, 50x50mm, thickness 0.009mm, 99.9%
Tantalum, foil, not light tested, 50x50mm, thickness 0.004mm, 99.9%
Tantalum, foil, not light tested, 100x100mm, thickness 0.0025mm, 99.9%
Tantalum, foil, 10m coil, thickness 0.04mm, coil width 0.7mm, annealed, 99.9%
Tantalum, foil, 10mm disks, thickness 0.034mm, as rolled, 99.9%
Tantalum, tube, 100mm, outside diameter 0.80mm, inside diameter 0.50mm, wall thickness 0.15mm, as drawn, 99.9%
Tantalum, tube, 200mm, outside diameter 3.1mm, inside diameter 2.6mm, wall thickness 0.25mm, as drawn, 99.9%
Tantalum, wire reel, 0.5m, diameter 0.8mm, annealed, 99.9%
Tantalum, tube, 500mm, outside diameter 1.02mm, inside diameter 0.86mm, wall thickness 0.08mm, as drawn, 99.9%
Tantalum, tube, 100mm, outside diameter 6.35mm, inside diameter 4.35mm, wall thickness 1mm, as drawn, 99.9%
Tantalum, tube, 500mm, outside diameter 2.0mm, inside diameter 1.4mm, wall thickness 0.3mm, as drawn, 99.9%
Tantalum, wire reel, 20m, diameter 0.25mm, annealed, 99.9%
Tantalum, wire reel, 10m, diameter 0.25mm, annealed, 99.9%
Tantalum, tube, 1000mm, outside diameter 3.1mm, inside diameter 2.6mm, wall thickness 0.25mm, as drawn, 99.9%
Tantalum, tube, 200mm, outside diameter 10.0mm, inside diameter 8.4mm, wall thickness 0.8mm, as drawn, 99.9%
Tantalum, wire reel, 5m, diameter 0.025mm, as drawn, 99.9%
Tantalum, wire reel, 5m, diameter 0.40mm, annealed, 99.9%
Tantalum, wire reel, 0.5m, diameter 1.0mm, annealed, 99.9%
Tantalum, tube, 500mm, outside diameter 2.0mm, inside diameter 1mm, wall thickness 0.5mm, as drawn, 99.9%
Tantalum, tube, 200mm, outside diameter 2.0mm, inside diameter 1.4mm, wall thickness 0.3mm, as drawn, 99.9%
Tantalum, tube, 100mm, outside diameter 9.5mm, inside diameter 8.5mm, wall thickness 0.5mm, as drawn, 99.9%
Tantalum, wire reel, 100m, diameter 0.125mm, annealed, 99.9%
Tantalum, wire reel, 50m, diameter 0.25mm, annealed, 99.9%
Tantalum, tube, 500mm, outside diameter 3.5mm, inside diameter 3.1mm, wall thickness 0.2mm, as drawn, 99.9%
Sigma-Aldrich
Tantalum, powder, 99.99% trace metals basis
Sigma-Aldrich
Tantalum, foil, thickness 0.5 mm, ≥99.9% trace metals basis
Sigma-Aldrich
Tantalum, foil, thickness 0.25 mm, ≥99.9% trace metals basis
Sigma-Aldrich
Tantalum, foil, thickness 1.0 mm, ≥99.9% trace metals basis
Sigma-Aldrich
Tantalum, foil, thickness 0.025 mm, ≥99.9% trace metals basis