Merck
Advances in identifying beryllium sensitization and disease.
International journal of environmental research and public health (2010-03-03)
Dan Middleton, Peter Kowalski
ABSTRACT
Beryllium is a lightweight metal with unique qualities related to stiffness, corrosion resistance, and conductivity. While there are many useful applications, researchers in the 1930s and 1940s linked beryllium exposure to a progressive occupational lung disease. Acute beryllium disease is a pulmonary irritant response to high exposure levels, whereas chronic beryllium disease (CBD) typically results from a hypersensitivity response to lower exposure levels. A blood test, the beryllium lymphocyte proliferation test (BeLPT), was an important advance in identifying individuals who are sensitized to beryllium (BeS) and thus at risk for developing CBD. While there is no true "gold standard" for BeS, basic epidemiologic concepts have been used to advance our understanding of the different screening algorithms.
Product Description
Beryllium, wire reel, 0.05m, diameter 0.75mm, annealed and clean, 99%
Beryllium, wire reel, 0.1m, diameter 0.05mm, annealed and clean, 99.7%
Beryllium, wire reel, 0.2m, diameter 0.356mm, annealed and clean, 99.7%
Beryllium, wire reel, 0.2m, diameter 0.125mm, annealed and clean, 99.7%
Beryllium, wire reel, 0.05m, diameter 0.5mm, annealed and clean, 99%
Beryllium, wire reel, 0.2m, diameter 0.5mm, annealed and clean, 99%
Beryllium, wire reel, 0.1m, diameter 0.125mm, annealed and clean, 99.7%
Beryllium, wire reel, 0.1m, diameter 0.25mm, annealed and clean, 99.7%
Beryllium, wire reel, 0.1m, diameter 0.356mm, annealed and clean, 99.7%
Beryllium, wire reel, 0.05m, diameter 0.25mm, annealed and clean, 99.7%
Beryllium, wire reel, 0.2m, diameter 0.75mm, annealed and clean, 99%
Beryllium, wire reel, 0.1m, diameter 0.5mm, annealed and clean, 99%
Beryllium, wire reel, 0.2m, diameter 0.85mm, annealed and clean, 99%
Beryllium, wire reel, 0.05m, diameter 0.356mm, annealed and clean, 99.7%
Beryllium, wire reel, 0.1m, diameter 0.6mm, annealed and clean, 99%
Beryllium, wire reel, 0.1m, diameter 0.75mm, annealed and clean, 99%
Beryllium, wire reel, 0.05m, diameter 0.6mm, annealed and clean, 99%
Beryllium, wire reel, 0.05m, diameter 0.85mm, annealed and clean, 99%
Beryllium, wire reel, 0.1m, diameter 0.85mm, annealed and clean, 99%
Beryllium, wire reel, 0.2m, diameter 0.25mm, annealed and clean, 99.7%
Beryllium, wire reel, 0.2m, diameter 0.6mm, annealed and clean, 99%
Beryllium, wire reel, 5m, diameter 0.05mm, annealed and clean, 99.7%
Beryllium, wire reel, 1m, diameter 0.25mm, annealed and clean, 99.7%
Beryllium, wire reel, 0.5m, diameter 0.125mm, annealed and clean, 99.7%
Beryllium, wire reel, 0.3m, diameter 0.356mm, annealed and clean, 99.7%
Beryllium, wire reel, 0.5m, diameter 0.356mm, annealed and clean, 99.7%
Beryllium, wire reel, 0.5m, diameter 0.05mm, annealed and clean, 99.7%
Beryllium, wire reel, 0.5m, diameter 0.25mm, annealed and clean, 99.7%
Beryllium, wire reel, 1m, diameter 0.05mm, annealed and clean, 99.7%
Beryllium, wire reel, 1m, diameter 0.125mm, annealed and clean, 99.7%