Merck
  • Application of the margin of exposure (MoE) approach to substances in food that are genotoxic and carcinogenic: example: aflatoxin B1 (AFB1).
Application of the margin of exposure (MoE) approach to substances in food that are genotoxic and carcinogenic: example: aflatoxin B1 (AFB1).
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association (2010-02-02)
Diane Benford, Jean-Charles Leblanc, R Woodrow Setzer
ABSTRACT
Aflatoxin B1 (AFB1) has been consistently shown to be a potent mutagen and is a liver carcinogen in humans and a wide range of animal species. The Fisher rat appears to be the most sensitive animal model. Dose-response modelling results in a BMDL(10) of 250 ng/kg-bw/d. Estimates of mean dietary exposure estimates were 0.4 ng/kg-bw/d and 2.6 ng/kg-bw/d, representing regions with low and high exposure, respectively. The MOEs for mean exposure therefore range from 100 to 600.
Product Description
Supelco
Aflatoxin B1 solution, 3 μg/mL in benzene:acetonitrile (98:2), analytical standard
Aflatoxin B1 solution, 3.79 μg/g in acetonitrile, ERM®, certified reference material
Supelco
Aflatoxin B1 solution, 2 μg/mL in acetonitrile, analytical standard
Sigma-Aldrich
Aflatoxin B1 from Aspergillus flavus, from Aspergillus flavus