Merck
  • [Comparative analysis of sensitivity of proteases (chymotrypsin and trypsin) and cholinesterases of different origin to some organophosphorus inhibitors].
[Comparative analysis of sensitivity of proteases (chymotrypsin and trypsin) and cholinesterases of different origin to some organophosphorus inhibitors].
Zhurnal evoliutsionnoi biokhimii i fiziologii (2009-07-03)
E V Rozengart
ABSTRACT
The antichymotrypsin, antitrypsin, and anticholinesterase efficiencies of four homologous series of organophosphorus inhibitors are compared: O-ethyl-S-(n-alkyl)methylthiophosphonates, O-(n-alkyl)-S-(n-butyl)methylthiophosphonates, O-(n-alkyl)-S-beta-(ethylmercaptoethylene)methylthiophosphonates, and their methylsulfomethylates. As sources of a-chymotrypsin and trypsin, commercial compounds of Worthington Biochemical Corporation and Leningrad Myasokombinat were tested. Bimolecular constant of the reaction rate was used as the measure of antienzyme efficiency. In all cases, the antichymotrypsin efficiency was lower, while the antitrypsin--essentially higher than the anticholinesterase activity of the studied inhibitors. These differences were found to much depend both on the inhibitor structure and on nature of the cholinesterase compounds.
Product Description
Chymotrypsin, BRP, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
α-Chymotrypsin from bovine pancreas, ≥40 units/mg protein, vial of 5 mg
Sigma-Aldrich
α-Chymotrypsin from bovine pancreas, Type II, lyophilized powder, ≥40 units/mg protein
Sigma-Aldrich
α-Chymotrypsin from bovine pancreas, (TLCK treated to inactivate residual tryspin activity), Type VII, essentially salt-free, lyophilized powder, ≥40 units/mg protein
Sigma-Aldrich
α-Chymotrypsin from bovine pancreas, suitable for protein sequencing, salt-free, lyophilized powder
Sigma-Aldrich
α-Chymotrypsin from bovine pancreas, Type I-S, essentially salt-free, lyophilized powder