Merck
Sabouraud('s) agar.
Journal of medical and veterinary mycology : bi-monthly publication of the International Society for Human and Animal Mycology (1991-01-01)
F C Odds
ABSTRACT
'Sabouraud glucose agar' is the name recommended for present-day versions of the medium originally designed by Raymond Sabouraud. The title 'Sabouraud glucose agar' implies a medium comprising 1% (w:v) peptone, 4% (w:v) glucose and 1.5-2.0% (w:v) agar with a final pH of 5.6 and should not be applied to any other formulation, in the interests of inter-laboratory reproducibility. Use of the word 'dextrose' as a synonym for 'glucose' is not recommended. The colony forms, microscopic characteristics and other physiological properties of fungi cultivated on Sabouraud glucose agar may vary considerably according to the type of peptone used in the medium. The type of agar chosen and the conditions of autoclave sterilization are further obstacles to true standardization of this widely used mycological culture medium.
Product Description
Millipore
Agar, suitable for microbiology
Millipore
Agar, suitable for microbiology
Sigma-Aldrich
Noble agar
Sigma-Aldrich
Agar, High gel strength, suitable for plant cell culture
Sigma-Aldrich
Agar, suitable for plant cell culture, powder
Sigma-Aldrich
Agar, Type A, suitable for plant cell culture
Sigma-Aldrich
Select agar
Sigma-Aldrich
Agar, suitable for plant cell culture
Sigma-Aldrich
Agar, Type M, suitable for plant cell culture
Sigma-Aldrich
Agar, microbiology tested, suitable for plant cell culture, suitable for cell culture, powder
Sigma-Aldrich
Agar, Bacteriological, microbiology tested, suitable for plant cell culture, granular
Sigma-Aldrich
Agar, ash 2.0-4.5%
Millipore
Agar, powder, suitable for microbiology
Millipore
Agar, high purity, low ionic content, low gel strenght, suitable for microbiology
Sigma-Aldrich
Bacteriological agar, for molecular biology