Merck
  • Single-dose azithromycin versus erythromycin or amoxicillin for Chlamydia trachomatis infection during pregnancy: a meta-analysis of randomised controlled trials.
Single-dose azithromycin versus erythromycin or amoxicillin for Chlamydia trachomatis infection during pregnancy: a meta-analysis of randomised controlled trials.
International journal of antimicrobial agents (2007-06-29)
Eleni Pitsouni, Christos Iavazzo, Stavros Athanasiou, Matthew E Falagas
ABSTRACT
This review aimed to compare data regarding the effectiveness and safety of azithromycin with alternative regimens in the treatment of pregnant women with Chlamydia trachomatis infection. PubMed and Scopus databases were searched to identify relevant randomised controlled trials (RCTs). The main analysis focused on comparison of azithromycin with erythromycin. In a secondary analysis, azithromycin was compared with erythromycin or amoxicillin. Eight RCTs studying 587 pregnant women with microbiologically documented C. trachomatis infection were included in the meta-analysis. Overall, there was no difference between azithromycin and erythromycin regarding treatment success in intention-to-treat patients (pooled odds ratio (OR)=2.66, 95% confidence interval (CI) 0.69-10.29) or in clinically evaluated patients (OR=1.46, 95% CI 0.56-3.78). Furthermore, azithromycin was associated with fewer gastrointestinal adverse events (OR=0.11, 95% CI 0.07-0.18), fewer total adverse events (OR=0.11, 95% CI 0.07-0.18), a smaller proportion of patients who withdrew from the study (OR=0.12, 95% CI 0.04-0.37) and better compliance (OR=23.7, 95% CI 9.34-60.14) than erythromycin. The results of the secondary analysis comparing azithromycin with erythromycin or amoxicillin were similar to those of the main analysis. In conclusion, azithromycin was associated with similar effectiveness but less adverse events compared with erythromycin or amoxicillin in the treatment of pregnant women with C. trachomatis infection.
Sigma-Aldrich
Amoxicillin, 95.0-102.0% anhydrous basis
Supelco
Erythromycin, Pharmaceutical Secondary Standard; Certified Reference Material
Erythromycin, for microbiological assay, European Pharmacopoeia (EP) Reference Standard
USP
Erythromycin, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Erythromycin, meets USP testing specifications
Sigma-Aldrich
Erythromycin, BioReagent, suitable for cell culture
Sigma-Aldrich
Erythromycin, potency: ≥850 μg per mg
Sigma-Aldrich
Erythromycin, tested according to Ph. Eur.
Sigma-Aldrich
Erythromycin standard solution, 1 mg/mL in H2O