Merck
Holmium laser for stone management.
World journal of urology (2007-03-07)
Sean Pierre, Glenn M Preminger
ABSTRACT
The efficiency and safety profile of the holmium laser has made this tool a versatile multi-purpose instrument for use in the endoscopic treatment of a wide variety of urologic disorders, in particular urinary calculi. Herein we review holmium laser physics, current endourologic laser lithotripsy applications, and the performance of new low power holmium laser devices.
Sigma-Aldrich
Holmium, chips, 99.9% trace metals basis
Holmium, rod, 100mm, diameter 3.8mm, 99%
Holmium, foil, 50x50mm, thickness 0.125mm, as rolled, 99%
Holmium, foil, not light tested, 25x25mm, thickness 0.0125mm, as rolled, 99%
Holmium, rod, 100mm, diameter 6.35mm, cast, 99%
Holmium, foil, not light tested, 25x25mm, thickness 0.005mm, as rolled, 99%
Holmium, rod, 200mm, diameter 3.8mm, 99%
Holmium, foil, not light tested, 50x50mm, thickness 0.0125mm, as rolled, 99%
Holmium, foil, not light tested, 25x25mm, thickness 0.025mm, as rolled, 99%
Holmium, foil, not light tested, 50x50mm, thickness 0.025mm, as rolled, 99%
Holmium, rod, 50mm, diameter 6.35mm, cast, 99%
Holmium, foil, not light tested, 100x100mm, thickness 0.0125mm, as rolled, 99%
Holmium, rod, 50mm, diameter 3.8mm, 99%
Holmium, foil, light tested, 50x50mm, thickness 0.05mm, as rolled, 99%
Holmium, foil, 25x25mm, thickness 0.125mm, as rolled, 99%
Holmium, foil, light tested, 25x25mm, thickness 0.05mm, as rolled, 99%
Holmium, powder, 10g, max. particle size 500 micron, 99.9%