Merck
  • Tobramycin solution for inhalation in cystic fibrosis patients: a review of the literature.
Tobramycin solution for inhalation in cystic fibrosis patients: a review of the literature.
Expert opinion on pharmacotherapy (2007-02-21)
Jennifer K Hagerman, Stephanie A Knechtel, Michael E Klepser
ABSTRACT
Cystic fibrosis patients suffer increased sputum production and a notable decline in respiratory function throughout the progression of their disease. Patients are left vulnerable to respiratory colonization/infection from a number of pathogens, including Pseudomonas aeruginosa. At present, the only antibiotic formulation that is FDA approved for aerosolized delivery is tobramycin solution for inhalation (TSI). TSI allows for targeted antibiotic delivery to the lungs and is indicated for maintenance therapy in cystic fibrosis patients infected with P. aeruginosa. Studies demonstrate that cyclical treatment with TSI is associated with an increase in respiratory function and a decrease in sputum density in cystic fibrosis patients. Additional benefits include fewer hospitalizations and a decreased need for systemic antibiotics. However, because of the need for chronic administration, issues such as emergence of resistant organisms and toxicity are a potential concern and have also been evaluated. This review details the pharmacology of TSI and literature involving its use in cystic fibrosis patients.
Sigma-Aldrich
Tobramycin, Aminoglycoside antibiotic
Tobramycin, European Pharmacopoeia (EP) Reference Standard
Tobramycin for identification, European Pharmacopoeia (EP) Reference Standard
Supelco
Tobramycin, Pharmaceutical Secondary Standard; Certified Reference Material