Merck
  • [Itraconazole in the treatment of superficial candidal infections: twelve years' clinical experience].
[Itraconazole in the treatment of superficial candidal infections: twelve years' clinical experience].
Zhongguo yi xue ke xue yuan xue bao. Acta Academiae Medicinae Sinicae (2006-08-12)
Shi-Hai Xie, An-Sheng Li, Peng-Cheng Ma, Zhi-Qiang Chen
ABSTRACT
Itraconazole has been used to treat superficial candidal infections in China for 12 years with promising efficacy and safety. This article retrospectively reviewed literatures published in the mainstream journals in China with an attempt to find a reasonable therapy for Chinese populations.
Product Description
Itraconazole, European Pharmacopoeia (EP) Reference Standard
Itraconazole for system suitability, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Itraconazole, ≥98% (HPLC)