Merck
Lindane-induced liver oxidative stress.
Free radical biology & medicine (1990-01-01)
L A Videla, S B Barros, V B Junqueira
ABSTRACT
The development of an oxidative stress condition in the liver by lindane intoxication is discussed as a possible hepatotoxic mechanism of the insecticide. Lindane is metabolized by liver microsomal enzymes to a variety of metabolites, which are susceptible of conjugation for proper elimination. In addition, the interaction of lindane with the liver tissue results in the induction of the microsomal cytochrome P-450 system, together with enhanced rates of superoxide radical generation and a significant increase in indicators of lipid peroxidation. Concomitantly, lindane intoxication induces a derangement of some antioxidant mechanisms of the liver cell, including decreased superoxide dismutase and catalase activities and alterations in reduced glutathione content leading to depressed GSH/GSSG ratios. The time course study of the changes in hepatic lipid peroxidation and antioxidant parameters are closely interrelated and coincide with the onset and progression of morphological lesions.