Merck
Aflatoxin B1-induced DNA damage and its repair.
Cancer letters (2006-02-07)
Leanne L Bedard, Thomas E Massey
ABSTRACT
Aflatoxin B(1) (AFB(1))-N(7)-guanine is the predominant adduct formed upon the reaction of AFB(1)-8,9-exo-epoxide with guanine residues in DNA. AFB(1)-N(7)-guanine can convert to the ring-opened formamidopyrimidine, or the adducted strand can undergo depurination. AFB(1)-N(7)-guanine and AFB(1)-formamidopyrimidine are thought to be predominantly repaired by nucleotide excision repair in bacteria, yeast and mammals. Although AFB(1)-formamidopyrimidine is removed less efficiently than AFB(1)-N(7)-guanine in mammals, both lesions are repaired with equal efficiencies in bacteria, reflecting differences in damage recognition between bacterial and mammalian repair systems. Furthermore, DNA repair activity and modulation of repair by AFB(1) seem to be major determinants of susceptibility to AFB(1)-induced carcinogenesis.
Sigma-Aldrich
Aflatoxin B1 from Aspergillus flavus, from Aspergillus flavus
Aflatoxin B1 solution, 3.79 μg/g in acetonitrile, ERM®, certified reference material
Supelco
Aflatoxin B1 solution, 2 μg/mL in acetonitrile, analytical standard
Supelco
Aflatoxin B1 solution, 3 μg/mL in benzene:acetonitrile (98:2), analytical standard