Merck
  • Promising treatment of autoimmune hepatitis with 6-thioguanine after adverse events on azathioprine.
Promising treatment of autoimmune hepatitis with 6-thioguanine after adverse events on azathioprine.
European journal of gastroenterology & hepatology (2005-03-10)
Nanne K H de Boer, Carin M J van Nieuwkerk, M Nieves Aparicio Pages, Sybrand Y de Boer, Luc J J Derijks, Chris J J Mulder
ABSTRACT
The use of corticosteroids in autoimmune hepatitis is an established therapy. To avoid the possible serious side effects of corticosteroids, immunosuppression with azathioprine is often warranted. Azathioprine, a purine analogue, is frequently used to taper or replace corticosteroids. However, approximately 10% of the patients are intolerant to azathioprine. Alternative therapies using mycophenolate, tacrolimus, budesonide, cyclosporine and 6-mercaptopurine have been studied, with variable results. The use of 6-thioguanine, an agent more directly leading to the down-stream active metabolites of azathioprine (6-thioguanine nucleotides) in inflammatory bowel disease patients intolerant to azathioprine or 6-mercaptopurine showed conflicting results. We report three patients with autoimmune hepatitis who could not tolerate azathioprine but tolerated 6-thioguanine 0.3 milligram per kilogram daily well. All three patients improved clinically. Therapeutic drug monitoring was performed. The prospective evaluation of 6-thioguanine as a possible immunosuppressive drug in autoimmune hepatitis patients is warranted.
Sigma-Aldrich
6-Thioguanine, ≥98%
Sigma-Aldrich
6-Thioguanine, Hybri-Max, 50 ×, γ-irradiated, lyophilized powder, BioXtra, suitable for hybridoma