Merck
  • Meta-analysis of randomised controlled prostaglandin E1 studies in peripheral arterial occlusive disease stages III and IV.
Meta-analysis of randomised controlled prostaglandin E1 studies in peripheral arterial occlusive disease stages III and IV.
VASA. Zeitschrift fur Gefasskrankheiten (2004-10-06)
A Creutzig, W Lehmacher, M Elze
ABSTRACT
The relevance of Prostaglandin El (PGE,) in the treatment of peripheral arterial occlusive disease stage III and IV was to be evaluated for the first time by a meta-analysis. Altogether, 643 patients were analyzed from seven randomized, controlled PGE1 studies that were comparable with regard to patient selection, study design and endpoints. Of these, only placebo-controlled studies (n = 254) were included in the formal meta-analysis using the method of DerSimonian and Laird. Additionally, the response rate and the rate of adverse events were determined for the pooled groups of all studies. At the end of treatment, PGE1 showed a significantly better response (ulcer healing and/or pain reduction) as compared to placebo (47.8% for PGE1, vs. 25.2% for placebo, p = 0.0294). A significant difference in favor of PGE1 was also seen for the combined endpoint "major amputation or death" after 6-month follow-up (22.6% for PGE1 vs. 36.2% for placebo, p = 0.0150). The response rate (ulcer healing and/or pain relief) of the pooled treatment groups was 60.2% for PGE1, 25.2% for placebo, and 53.6% for iloprost. The adverse events rate of the pooled treatment groups showed good tolerability for PGE, with a rate of 39.6% in comparison to 73.9% for iloprost and 15.4% for placebo. For patients with peripheral arterial occlusive disease stage III or IV not eligible for arterial reconstruction, PGE1 therapy not only has significant beneficial effects over placebo on ulcer healing and pain relief but also increases the rate of patients surviving with both legs after 6-months follow-up.
Sigma-Aldrich
Prostaglandin E1, synthetic, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Prostaglandin E1, ≥98% (HPLC), synthetic
Sigma-Aldrich
Prostaglandin E1, γ-irradiated, powder, BioXtra, suitable for cell culture
Alprostadil, European Pharmacopoeia (EP) Reference Standard